Supelco® Organic Reference Standard, (−)-Carvone
(−)-Carvone, Analytical standard, Glass bottle, 5 mL
Katalog # 22060-5ML-F
Leverandør: Merck
Gå til:
Product Details & Documents
- GHS signal: GHS07
Advarsel: P261 - P264 - P270 - P280 - P301 + P312 - P302 + P352
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL
- Standard:Analytical standard
- Beskrivelse:(−)-Carvone
- Densitet:0,960 g/ml at 20 °C (lit.)
- Cat. No.:22060-5ML-F
- M-Clarity quality level:MQ100
- CAS-nr:6485-40-1
- Molekylvægt:150,22
- MDL No.:MFCD00001578
- Kogepunkt:228...230 °C (lit.)
- Fareklassebeskrivelse:H302 - H317
- Flammepunkt:89 °C - closed cup
- Opbevaring betingelser:2...30 °C
- Synonym(s):(−)-p-Mentha 6,8-diene 2-one; (R)-5-Isopropenyl-2-methyl-2-cyclohexenone; Carvol
- Shipping conditions:STD
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



