Orders LinkContactUsLinkComponent
Tangential flow cassettes, Centramate™
Tangential flow cassettes, Centramate™
Supor®, 0,1 µm, 1000 cm²
Katalog # 516-9693
Tangential flow cassettes, Centramate™
Katalog # 516-9693
Tangential flow cassettes, Centramate™
Tangential flow cassettes, Centramate™
Supor®, 0,1 µm, 1000 cm²
Price
13.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Filtreringsområde:
    1000 cm²
  • Membran:
    Supor®
  • Porestørrelse:
    0,1 µm
  • Seq:
    03