Orders LinkContactUsLinkComponent
Pasteur pipettes, Corning®
Pasteur pipettes, Corning®
Leverandør:   Corning
Pasteur pipettes, Corning®
Leverandør:  Corning
Certifikater
Pasteur pipettes, Corning®
Pasteur pipettes, Corning®
3 tilgængelige muligheder nedenfor
Product Details & Documents
Soda-lime or borosilicate glass.
  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, haematology and serology studies, as well as blood bank and clinical chemistry procedures.
Product Family Options
Produktinformation
  • Udv.Ø
    Længde
    Steril
    Pakket
    Tilgængelighed
    Price
  • 612-5199
    Non plugged, borosilicate glass
    Non plugged, borosilicate glass
    6,5 mm
    146 mm
    -
    5/pakke
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 641,00 kr
    641,00 kr
    1 * 1.000 Items
       641,00 kr
  • 612-5200
    Non plugged, borosilicate glass
    Non plugged, borosilicate glass
    6,5 mm
    229 mm
    -
    5/pakke
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 714,00 kr
    714,00 kr
    1 * 1.000 Items
       714,00 kr
  • 612-5198
    Non plugged, long tip, soda-lime glass
    Non plugged, long tip, soda-lime glass
    6,5 mm
    330 mm
    -
    100/kasse
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 998,00 kr
    998,00 kr
    1 * 100 Items
       998,00 kr