Svovlsyre concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32053.602
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pakning:Plastic ampoule
- Pakn:170 mL
- Seq:2515
Specifications
Specification Test Results
- Titer (20°C):0.4975 - 0.5025 mol/l
Specification Test Results
About this item
:
:98.08 g/mol
ADR:8,II
:Dosis til fremstilling af 1 L opløsning
EINECS:231-639-5
UN:2796
:GHS05
Storage Temperature:Stue temperatur
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589