Orders LinkContactUsLinkComponent
(±)-Epichlorhydrin, Supelco®
(±)-Epichlorhydrin, Supelco®
Glass bottle, 1 mL
Katalog # 02578-1ML
Brandfarlig
Giftig
Ætsende
Sundhed
Leverandør:   Merck
(±)-Epichlorhydrin, Supelco®
Katalog # 02578-1ML
Leverandør:  Merck
CAS nummer:  106-89-8
(±)-Epichlorhydrin, Supelco®
(±)-Epichlorhydrin, Supelco®
Glass bottle, 1 mL
CAS nummer:  106-89-8
Price
382,00 kr
Log ind for tilgængelighed
1 * 1 mL
Product Details & Documents
Synonymer:  Epichlorhydrin, 1-Chlor-2,3-epoxypropan, (±)-1-Chlor-2,3-epoxypropan, 3-Chlor-1,2-epoxypropan, (±)-3-Chlor-1,2-epoxypropan
  • CAS nummer:106-89-8
  • Formel:C₃H₅ClO
  • MDL nummer:MFCD00005132
  • Molecular Weight:92.52 g/mol
  • Merck Index:13,03642
  • Kogende Pt:117,9 °C (1013 hPa)
  • Smeltende Pt:–57,2 °C
  • Tæthed:0,9985 g/cm³ (20 °C)
  • Flash Pt:28 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:2023
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-439-8
Documents
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle
  • Pakn:
    1 mL