Natriumhydrogencarbonat-[13C] (99% ¹³C )
:87081-58-1
:
:NaHCO₃
:85 g/mol
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
VWR®, Nitril handsker, NITRILE
: VWR Collection
Vand, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Xylen ≥98%, TECHNICAL
: VWR Chemicals
Stopcock, three way, BD Connecta™
: BD Medical