Cycloocten, Supelco®
Glass bottle, 1 mL
Catalog # 29648-1ML
Supplier: Merck
Jump to:
Product Details & Documents
- CAS Number:931-88-4
- Formula:C₈H₁₄
- MDL Number:MFCD00001753
- Molecular Weight:110.2 g/mol
- Boiling Pt:146 °C (1013 hPa)
- Melting Pt:–16 °C
- Density:0,855 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:213-245-5
- UN:3295
- ADR:3,III
- :M-Clarity™ quality level = MQ100
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:1 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



