Multi-parameter meter, handheld, SevenGo Duo pro™ SG98
Om denne vare
Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and dissolved oxygen (optical RDO® technology). Ideal for special analyses, particularly in environmental, chemical and food and beverage industry.
- Intuitive, user friendly operation using menu guidance in ten languages and backlit display
- Fast data transmission thanks to wireless IR communication interface
- Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID
- Manual or automatic air pressure compensation for reliable results
- Optical oxygen sensor based on RDO® technology; reliable 'Intelligent Sensor Management' (ISM®)
- Rugged watertight housing (IP 67)
Electrode Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m cable); Electrode Kit with SG98 also includes InLab® Expert Pro ISM®.
Field Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m or 5 m cable), compact field case, rubber holster and OptiOx™ protective guard; Field Kit with SG98 also includes InLab® Expert Pro ISM®.
Delivery information: Meter only is supplied without probes.
The Electrode Kit includes the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 m cable).
The Field Kits include the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 or 5 m cable), OptiOx protective guard, rubber holster, calibration sachets, sample bottles and Field Compact Case.
4 tilgængelige muligheder nedenfor
Product Details & Documents
Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and dissolved oxygen (optical RDO® technology). Ideal for special analyses, particularly in environmental, chemical and food and beverage industry.
- Intuitive, user friendly operation using menu guidance in ten languages and backlit display
- Fast data transmission thanks to wireless IR communication interface
- Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID
- Manual or automatic air pressure compensation for reliable results
- Optical oxygen sensor based on RDO® technology; reliable 'Intelligent Sensor Management' (ISM®)
- Rugged watertight housing (IP 67)
Electrode Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m cable); Electrode Kit with SG98 also includes InLab® Expert Pro ISM®.
Field Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m or 5 m cable), compact field case, rubber holster and OptiOx™ protective guard; Field Kit with SG98 also includes InLab® Expert Pro ISM®.
Delivery information: Meter only is supplied without probes.
The Electrode Kit includes the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 m cable).
The Field Kits include the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 or 5 m cable), OptiOx protective guard, rubber holster, calibration sachets, sample bottles and Field Compact Case.
Documents
Specifications for Product Family
Specifikationer
- Catalog No:
- 665-0239
- 665-0237
- 665-0238
- 664-0164
- Model:
- SG98
- SG98
- SG98
- SG98
- pH - Område:
- –2,000...+19,999
- –2,000...+19,999
- –2,000...+19,999
- –2,000...+19,999
- pH - Aflæsningsinterval:
- 0,1/0,01/0,001
- 0,1/0,01/0,001
- 0,1/0,01/0,001
- 0,1/0,01/0,001
- pH - Nøjagtighed:
- ±0,002
- ±0,002
- ±0,002
- ±0,002
- pH - Kalibrering:
- Max. 5 points, 7 predefined buffer groups
- Max. 5 points, 7 predefined buffer groups
- Max. 5 points, 7 predefined buffer groups
- Max. 5 points, 7 predefined buffer groups
- mV - Område:
- ±1999,9
- ±1999,9
- ±1999,9
- ±1999,9
- mV - Aflæsningsinterval:
- 0,1
- 0,1
- 0,1
- 0,1
- mV - Nøjagtighed:
- ±0,1
- ±0,1
- ±0,1
- ±0,1
- ISE Område:
- 1,00-9 - 9,99+9
- 1,00-9 - 9,99+9
- 1,00-9 - 9,99+9
- 1,00-9 - 9,99+9
- ISE Nøjagtighed:
- ±0,5%
- ±0,5%
- ±0,5%
- ±0,5%
- ISE Enheder:
- mg/l, mmol/l, mol/l
- mg/l, mmol/l, mol/l
- mg/l, mmol/l, mol/l
- mg/l, mmol/l, mol/l
- DO Koncentration:
- 0,00 - 50,00 mg/L
- 0,00 - 50,00 mg/L
- 0,00 - 50,00 mg/L
- 0,00 - 50,00 mg/L
- D.O. - Aflæsningsinterval:
- 0,01 mg/L
- 0,01 mg/L
- 0,01 mg/L
- 0,01 mg/L
- D.O. - mætning:
- 0 - 500%
- 0 - 500%
- 0 - 500%
- 0 - 500%
- D.O. Nøjagtighed:
- ±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50) - ±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50) - ±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50) - ±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50)
- Temperaturinterval:
- DO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °C - DO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °C - DO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °C - DO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °C
- Temperature accuracy:
- DO: ±0,1 / pH: ±0,2 °C
- DO: ±0,1 / pH: ±0,2 °C
- DO: ±0,1 / pH: ±0,2 °C
- DO: ±0,1 / pH: ±0,2 °C
- Display:
- Dot matrix LCD
- Dot matrix LCD
- Dot matrix LCD
- Dot matrix LCD
- Strømforsyning:
- 4×1,5 V AA batteries or 1,3 V NiMH accumulators
- 4×1.5 V AA batteri eller 1.3 V NiMH akkumulator
- 4×1,5 V AA batteries or 1,3 V NiMH accumulators
- 4×1,5 V AA batteries or 1,3 V NiMH accumulators
- Overensstemmelse:
- IP 67/GLP
- IP 67/GLP
- IP 67/GLP
- IP 67/GLP
- Datalog Kapacitet:
- 500 measurements
- 500 data sæt
- 500 measurements
- 500 measurements
- Tilslutninger:
- IR to printer or PC via RS232 or USB
- IR til printer eller PC via RS232 eller USB
- IR to printer or PC via RS232 or USB
- IR to printer or PC via RS232 or USB
- Output:
- DO probe: Mini LTW
Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67) - DO probe: Mini LTW / Andre prober: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)
- DO probe: Mini LTW
Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67) - DO probe: Mini LTW
Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)
- B×D×H:
- 90×45×220 mm
- 90×45×220 mm
- 90×45×220 mm
- 90×45×220 mm
- Vægt:
- 0,368 kg
- 0,368 kg
- 0,368 kg
- 0,368 kg
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- 665-0239Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (5 m cable)Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (5 m cable)Log ind for tilgængelighed1 * 1 Items33.400,00 krSpecifikationer:
More
Specifications
Cat. No.665-0239ModelSG98pH - Område–2,000...+19,999pH - Aflæsningsinterval0,1/0,01/0,001pH - Nøjagtighed±0,002pH - KalibreringMax. 5 points, 7 predefined buffer groupsmV - Område±1999,9mV - Aflæsningsinterval0,1mV - Nøjagtighed±0,1ISE Område1,00-9 - 9,99+9ISE Nøjagtighed±0,5%ISE Enhedermg/l, mmol/l, mol/lDO Koncentration0,00 - 50,00 mg/LD.O. - Aflæsningsinterval0,01 mg/LD.O. - mætning0 - 500%D.O. Nøjagtighed±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50)TemperaturintervalDO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °CTemperature accuracyDO: ±0,1 / pH: ±0,2 °CDisplayDot matrix LCDStrømforsyning4×1,5 V AA batteries or 1,3 V NiMH accumulatorsOverensstemmelseIP 67/GLPDatalog Kapacitet500 measurementsTilslutningerIR to printer or PC via RS232 or USBOutputDO probe: Mini LTW
Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)B×D×H90×45×220 mmVægt0,368 kg - 665-0237Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Electrode KitHandheld multi-parameter meter, SevenGo Duo pro™ SG98, Electrode KitLog ind for tilgængelighed1 * 1 Items27.400,00 krSpecifikationer:
More
Specifications
Cat. No.665-0237ModelSG98pH - Område–2,000...+19,999pH - Aflæsningsinterval0,1/0,01/0,001pH - Nøjagtighed±0,002pH - KalibreringMax. 5 points, 7 predefined buffer groupsmV - Område±1999,9mV - Aflæsningsinterval0,1mV - Nøjagtighed±0,1ISE Område1,00-9 - 9,99+9ISE Nøjagtighed±0,5%ISE Enhedermg/l, mmol/l, mol/lDO Koncentration0,00 - 50,00 mg/LD.O. - Aflæsningsinterval0,01 mg/LD.O. - mætning0 - 500%D.O. Nøjagtighed±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50)TemperaturintervalDO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °CTemperature accuracyDO: ±0,1 / pH: ±0,2 °CDisplayDot matrix LCDStrømforsyning4×1.5 V AA batteri eller 1.3 V NiMH akkumulatorOverensstemmelseIP 67/GLPDatalog Kapacitet500 data sætTilslutningerIR til printer eller PC via RS232 eller USBOutputDO probe: Mini LTW / Andre prober: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)B×D×H90×45×220 mmVægt0,368 kg - 665-0238Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (1,8 m cable)Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (1,8 m cable)Log ind for tilgængelighed1 * 1 Items29.950,00 krSpecifikationer:
More
Specifications
Cat. No.665-0238ModelSG98pH - Område–2,000...+19,999pH - Aflæsningsinterval0,1/0,01/0,001pH - Nøjagtighed±0,002pH - KalibreringMax. 5 points, 7 predefined buffer groupsmV - Område±1999,9mV - Aflæsningsinterval0,1mV - Nøjagtighed±0,1ISE Område1,00-9 - 9,99+9ISE Nøjagtighed±0,5%ISE Enhedermg/l, mmol/l, mol/lDO Koncentration0,00 - 50,00 mg/LD.O. - Aflæsningsinterval0,01 mg/LD.O. - mætning0 - 500%D.O. Nøjagtighed±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50)TemperaturintervalDO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °CTemperature accuracyDO: ±0,1 / pH: ±0,2 °CDisplayDot matrix LCDStrømforsyning4×1,5 V AA batteries or 1,3 V NiMH accumulatorsOverensstemmelseIP 67/GLPDatalog Kapacitet500 measurementsTilslutningerIR to printer or PC via RS232 or USBOutputDO probe: Mini LTW
Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)B×D×H90×45×220 mmVægt0,368 kg - 664-0164Handheld multi-parameter meter, SevenGo Duo pro™ SG98, meter onlyHandheld multi-parameter meter, SevenGo Duo pro™ SG98, meter onlyLog ind for tilgængelighed1 * 1 Items18.040,00 krSpecifikationer:
More
Specifications
Cat. No.664-0164ModelSG98pH - Område–2,000...+19,999pH - Aflæsningsinterval0,1/0,01/0,001pH - Nøjagtighed±0,002pH - KalibreringMax. 5 points, 7 predefined buffer groupsmV - Område±1999,9mV - Aflæsningsinterval0,1mV - Nøjagtighed±0,1ISE Område1,00-9 - 9,99+9ISE Nøjagtighed±0,5%ISE Enhedermg/l, mmol/l, mol/lDO Koncentration0,00 - 50,00 mg/LD.O. - Aflæsningsinterval0,01 mg/LD.O. - mætning0 - 500%D.O. Nøjagtighed±0,1 mg/l (0 - 8)
±0,2 mg/l (8 - 20)
±10% (20 - 50)TemperaturintervalDO: 0...+50 °C
pH (ATC): –5...+130 °C
MTC: −30...+130 °CTemperature accuracyDO: ±0,1 / pH: ±0,2 °CDisplayDot matrix LCDStrømforsyning4×1,5 V AA batteries or 1,3 V NiMH accumulatorsOverensstemmelseIP 67/GLPDatalog Kapacitet500 measurementsTilslutningerIR to printer or PC via RS232 or USBOutputDO probe: Mini LTW
Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)B×D×H90×45×220 mmVægt0,368 kg
Accessories
Produktinformation
- Katalog #ModelKoncentrationTilgængelighedPrice
- Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67664-0076InLab® OptiOx™0 - 50 mg/l (0 - 500%)Log ind for tilgængelighed1 * 1 Items - 14.150,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- OptiOx™ beskyttelsesskærm
Produktinformation
- Katalog #TilgængelighedPrice
- OptiOx™ kalibreringsrør
Produktinformation
- Katalog #TilgængelighedPrice
- RDO udskiftningslåg til InLab® OptiOx™RDO udskiftningslåg til InLab® OptiOx™664-0078Log ind for tilgængelighed1 * 1 Items - 2.390,00 kr
Produktinformation
- Katalog #ModelKoncentrationTilgængelighedPrice
- Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67664-0077InLab® OptiOx™0 - 50 mg/l (0 - 500%)Log ind for tilgængelighed1 * 1 Items - 16.210,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- OptiOx™ BOD adapter
Produktinformation
- Katalog #ModelKoncentrationTilgængelighedPrice
- Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67664-0075InLab® OptiOx™0 - 50 mg/l (0 - 500%)Log ind for tilgængelighed1 * 1 Items - 14.780,00 kr