Jod 99.8+% ACS
Leverandør: Thermo Fisher Scientific
Om denne vare
- CAS nummer:7553-56-2
- Formel:I₂
- MDL nummer:MFCD00011355
- Molecular Weight:253.81 g/mol
- Merck Index:13,05036
- Kogende Pt:185 °C (760 mmHg)
- Smeltende Pt:113 °C
- Tæthed:4,9 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-442-4
- FN:3495
- ADR:8,III
3 tilgængelige muligheder nedenfor
Product Details & Documents
- CAS nummer:7553-56-2
- Formel:I₂
- MDL nummer:MFCD00011355
- Molecular Weight:253.81 g/mol
- Merck Index:13,05036
- Kogende Pt:185 °C (760 mmHg)
- Smeltende Pt:113 °C
- Tæthed:4,9 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-442-4
- FN:3495
- ADR:8,III
Documents
ACRO42382
SDS
Specifications for Product Family
Specifikationstestresultater
- Appearance:Grey Metallic spheres
- Titration Iodometric:≥99.8 %
- Non-volatile matter:≤0.01 %
- Chlorine and Bromine (as Cl):≤0.005 %