Orders LinkContactUsLinkComponent
Disposable spill kit
Disposable spill kit
Disposable spill kit
Katalog # 121-0136
Leverandør:   Brady
Disposable spill kit
Katalog # 121-0136
Leverandør:  Brady
Disposable spill kit
Disposable spill kit
Disposable spill kit
Price
736,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Designed for small spills of chemicals and hazardous liquids up to 1,7 litres. The highly visible dispenser box can easily be be wall or shelf mounted.
  • Disposable brush to sweep up any broken glass or debris
  • Protective gloves have roughened finger tips, increased tear resistance and good chemical resistance
  • Autoclavable disposal bag
 : Kit contains two absorbent pads (410×510 mm), one pair of disposable chemical protective gloves (powder free), disposal bag, one disposable brush, and multilingual instruction guide.
Specifikationer
  • Beskrivelse:
    Disposable spill kit