Orders LinkContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L
Catalog # AC10088
Supplier:   OMEGA BIO-TEK
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # AC10088
Supplier:  OMEGA BIO-TEK
CAS Number:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L
CAS Number:  
Price
575,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Specifications
Specifications
  • Pakning:
    Glass bottle
  • Pakn:
    1 L
  • Beskrivelse:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)