May Grunwald's eosin-methylenblåt, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
:
MDL Number:MFCD00131580
ADR:3,II
:M-Clarity™ quality level = MQ400
:GHS07
:GHS06
:GHS08
Storage Temperature:Stue temperatur
:1
Related Information
Merck The Color of Tissue Diagnostics
Product Literature
msds_101352
SDS
ProductPage_101352
Product specification
Frequently Bought Together