May Grunwald's eosin-methylenblåt opløsning, Sigma-Aldrich®
Catalog # MG80-2.5L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pakning:Glass bottle
- Pakn:2,5 L
- Seq:5096
Specifications
About this item
:
ADR:3,II
:M-Clarity™ quality level = MQ500
UN:1230
:GHS02
:GHS06
:GHS08
Storage Temperature:Stue temperatur
:1
MDL Number:MFCD00131580