Disulphineblå
Glass bottle for solids, 10 g
CHRO1B-447.0010
Marketsource
: CHROMA
: acid Blue1, Patent Blue, Patent Blue VF, Hydrogen [4-[4-(diethylamino)-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium, sodium salt, Patent blue VF sodium salt
- :129-17-9
- :C₂₇H₃₁N₂NaO₆S₂
- :MFCD00012119
- :566.67 g/mol
- :12,09161
- :290 °C
- :Stue temperatur
- :204-934-1
- Pakning:Glass bottle for solids
- Pakn:10 g