Orders LinkContactUsLinkComponent
Disulphineblå
Glass bottle for solids, 10 g
  CHRO1B-447.0010
Marketsource
 :   CHROMA
Disulphineblå
  CHRO1B-447.0010
 :  CHROMA
Marketsource
 :  129-17-9
Disulphineblå
Glass bottle for solids, 10 g
 :  129-17-9
1 * 10 g
 :  acid Blue1, Patent Blue, Patent Blue VF, Hydrogen [4-[4-(diethylamino)-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium, sodium salt, Patent blue VF sodium salt
  •  :129-17-9
  •  :C₂₇H₃₁N₂NaO₆S₂
  •  :MFCD00012119
  •  :566.67 g/mol
  •  :12,09161
  •  :290 °C
  •  :Stue temperatur
  •  :204-934-1
  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g