Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Menthol
Supelco® Organic Reference Standard, (−)-Menthol
Leverandør:   Merck
Supelco® Organic Reference Standard, (−)-Menthol
Leverandør:  Merck
Certifikater
Supelco® Organic Reference Standard, (−)-Menthol
Supelco® Organic Reference Standard, (−)-Menthol
2 tilgængelige muligheder nedenfor
Product Details & Documents
  • GHS signal: GHS07
Advarsel: P264 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313 - P337 + P313
Documents
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 63660-1G
  • 63660-100G
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • Synonym(s):
  • (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
  • (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
  • CAS-nr:
  • 2216-51-5
  • 2216-51-5
  • MDL No.:
  • MFCD00062979
  • MFCD00062979
  • Molekylvægt:
  • 156,27
  • 156,27
  • Melting point:
  • ~43 °C
  • ~43 °C
  • Fareklassebeskrivelse:
  • H315 - H319
  • H315 - H319
  • Flammepunkt:
  • 94 °C - closed cup
  • 94 °C - closed cup
  • Shipping conditions:
  • STD
  • STD
  • Opbevaring betingelser:
  • 2...30 °C
  • 2...30 °C
Specifikationer
Product Family Options
Produktinformation
  • Standard
    Pakning
    Pakn
    Tilgængelighed
    Price
  • (−)-Menthol
    Analytical standard
    Glass bottle for solids
    1 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 g
    1 * 1 g - 232,00 kr
    232,00 kr
    1 * 1 g
       232,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    63660-1G
    M-Clarity quality level
    MQ100
    Synonym(s)
    (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
    CAS-nr
    2216-51-5
    MDL No.
    MFCD00062979
    Molekylvægt
    156,27
    Melting point
    ~43 °C
    Fareklassebeskrivelse
    H315 - H319
    Flammepunkt
    94 °C - closed cup
    Shipping conditions
    STD
    Opbevaring betingelser
    2...30 °C
  • 63660-100G
    (−)-Menthol
    (−)-Menthol
    Analytical standard
    Glass bottle for solids
    100 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 g
    1 * 100 g - 534,00 kr
    534,00 kr
    1 * 100 g
       534,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    63660-100G
    M-Clarity quality level
    MQ100
    Synonym(s)
    (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
    CAS-nr
    2216-51-5
    MDL No.
    MFCD00062979
    Molekylvægt
    156,27
    Melting point
    ~43 °C
    Fareklassebeskrivelse
    H315 - H319
    Flammepunkt
    94 °C - closed cup
    Shipping conditions
    STD
    Opbevaring betingelser
    2...30 °C