TeleScoop
Om denne vare
TeleScoop is a versatile sampling system with exchangeable tools for a wide range of applications. The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint. Ideal for sampling from baths, tanks, shafts, open water courses. Sampling depths down to 6 m can be reached with the adjustable telescopic aluminium rod.
- For industry and water treatment
Angular beaker adaptable to different container shapes by adjustable angle setting (up to 90°).
PP pendulum beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 150 mm).
Stainless steel AISI 304 (1.4301) pendulum beaker automatically swinging to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 130 mm).
Bottle holder holds all bottles up to 88 mm Ø. Plastic, glass or metal bottles can be securely filled in this way. Angle steplessly adjustable up to 90°. With universal quick tightening strap. PP/PA.
Stainless steel AISI 304 (1.4301) beaker with scraper for powder, granules, pastes, mud. Sterilisable.
Stainless steel beaker V2A, extended rod with closed handle with immersion depth up to approx. 50 cm.
Surface and depth catch nets for taking solid and organic samples from water courses.
Telescopic rod: Different tools can be attached to the telescopic rod (snap-in joint). The individual rods have been provided with an end stop. This means that one rod cannot accidentally slip out of another rod. The telescopic rods cannot be rotated radially, which means even better handling. Increased telescopic rod rigidity by using cold-rolled profile rod. Springs made of stainless steel.
19 tilgængelige muligheder nedenfor
Product Details & Documents
TeleScoop is a versatile sampling system with exchangeable tools for a wide range of applications. The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint. Ideal for sampling from baths, tanks, shafts, open water courses. Sampling depths down to 6 m can be reached with the adjustable telescopic aluminium rod.
- For industry and water treatment
Angular beaker adaptable to different container shapes by adjustable angle setting (up to 90°).
PP pendulum beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 150 mm).
Stainless steel AISI 304 (1.4301) pendulum beaker automatically swinging to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 130 mm).
Bottle holder holds all bottles up to 88 mm Ø. Plastic, glass or metal bottles can be securely filled in this way. Angle steplessly adjustable up to 90°. With universal quick tightening strap. PP/PA.
Stainless steel AISI 304 (1.4301) beaker with scraper for powder, granules, pastes, mud. Sterilisable.
Stainless steel beaker V2A, extended rod with closed handle with immersion depth up to approx. 50 cm.
Surface and depth catch nets for taking solid and organic samples from water courses.
Telescopic rod: Different tools can be attached to the telescopic rod (snap-in joint). The individual rods have been provided with an end stop. This means that one rod cannot accidentally slip out of another rod. The telescopic rods cannot be rotated radially, which means even better handling. Increased telescopic rod rigidity by using cold-rolled profile rod. Springs made of stainless steel.
Product Family Options
Bægre med vinkelbeslag
Produktinformation
Bægre med pendulbeslag
Produktinformation
- MaterialeKapacitetTilgængelighedPrice
- 331-2120PendulbægerSpecifikationer:
More
Specifications
Cat. No.331-2120 - 331-0041PendulbægerSpecifikationer:
More
Specifications
Cat. No.331-0041 - 331-2121PendulbægerSpecifikationer:
More
Specifications
Cat. No.331-2121 - 331-2122PendulbægerSpecifikationer:
More
Specifications
Cat. No.331-2122
Flaskeholdere
Produktinformation
Bægre i rustfrit stål
Produktinformation
- MaterialeKapacitetTilgængelighedPrice
- 331-0040Bæger, rustfrit stålSpecifikationer:
More
Specifications
Cat. No.331-0040 - 129-1784TeleScoop, stainless steel beaker V2A, extended rodTeleScoop, stainless steel beaker V2A, extended rod1000 mlLog ind for tilgængelighed1 * 1 Items3.840,00 krSpecifikationer:
More
Specifications
Cat. No.129-1784
Opsamlingsnet
Produktinformation
- MaterialeKapacitetTilgængelighedPrice
- 300-0061Overfladenet, PVC/PA, blåt, 310 x 280 mm, maskestørrelse 2-5 mmOverfladenet, PVC/PA, blåt, 310 x 280 mm, maskestørrelse 2-5 mmLog ind for tilgængelighed1 * 1 Items730,00 krSpecifikationer:
More
Specifications
Cat. No.300-0061 - 331-0042Net, PP/PA, blå, 190 x 430 mm, netmasker 2-5 mmSpecifikationer:
More
Specifications
Cat. No.331-0042
Teleskopstænger
Produktinformation
- MaterialeKapacitetTilgængelighedPrice
- 300-0037TeleScoop, teleskopstang, 650 - 1200 mmSpecifikationer:
More
Specifications
Cat. No.300-0037 - 300-0237TeleScoop, telescopic rod, 700 - 1800 mmSpecifikationer:
More
Specifications
Cat. No.300-0237 - 300-0038Teleskopstang, 95-280 cmSpecifikationer:
More
Specifications
Cat. No.300-0038 - 300-0039TeleScoop, teleskopstang, 1150 - 3000 mmSpecifikationer:
More
Specifications
Cat. No.300-0039 - 300-0026Teleskopstang, 125-250 cmSpecifikationer:
More
Specifications
Cat. No.300-0026 - 331-2143TeleScoop, teleskopstav, 1650 - 4500 mmSpecifikationer:
More
Specifications
Cat. No.331-2143 - 300-0040Teleskopstang, 175-600 cmSpecifikationer:
More
Specifications
Cat. No.300-0040