Industrikøleskabe
Leverandør: Liebherr
Om denne vare
Commercial refrigerators with forced air cooling; the ventilation stops when door is opened.
- Automatic defrosting
- Lockable door
- LED interior lighting (glass models only)
- Interchangeable hinge
- One year warranty
Temperaturområde: 1 til 15 °C (FKv 503: 1 til 12 °C).
4 tilgængelige muligheder nedenfor
Product Details & Documents
Commercial refrigerators with forced air cooling; the ventilation stops when door is opened.
- Automatic defrosting
- Lockable door
- LED interior lighting (glass models only)
- Interchangeable hinge
- One year warranty
Temperaturområde: 1 til 15 °C (FKv 503: 1 til 12 °C).
Documents
FK4140-20
Datasheet
Specifications for Product Family
Specifikationer
- Catalog No:
- 471-0471
- LIEBFKUV1660
- 471-1064
- 471-0225
- 471-1034
- 472-0008
- 471-0410
- 471-1030
- 471-0230
- 471-1036
- 471-0693
- 471-1062
- 471-1180
- 471-1033
- 471-1031
- 471-1029
- 472-2183
- 472-1051
- 472-2184
- 472-2185
- 472-2195
- 472-2194
- 472-2191
- 472-2192
- 472-2250
- 472-2240
- 471-0231
- LIEBFKVSL5410
- 471-0232
- LIEBFKVSL2613
- 471-0234
- LIEBFKVSL3613
- LIEBFKVSL4112
- LIEBFKVSL5412
- LIEBFKVSL5413
- LIEBFK2640
- LIEBFK3640
- Model:
- FKUv 1610
- FKUv 1660
- FKv 1800
- FKv 2610
- FKv 2640
- FKv 4310
- FKv 5410
- FKv 5440
- FKv 502
- FKv 503
- FKUv 1612
- FKUv 1613
- FKUv 1663
- FKv 2643
- FKv 3643
- FKv 5443
- FKs 1800
- FKs 2600
- FKs 3600
- FKs 5000
- FKs 1802
- FKs 2602
- FKs 3602
- FKs 5002
- UKs 1801
- UKs 2602
- FKvsl 2610
- FKvsl 5410
- FKvsl 2612
- FKvsl 2613
- FKvsl 3612
- FKvsl 3613
- FKvsl 4112
- FKvsl 5412
- FKvsl 5413
- FK 2640
- FK 3640
- Art.nr:
- 471-0471
- LIEBFKUV1660
- 471-1064
- 471-0225
- 471-1034
- 472-0008
- 471-0410
- 471-1030
- 471-0230
- 471-1036
- 471-0693
- 471-1062
- 471-1180
- 471-1033
- 471-1031
- 471-1029
- 472-2183
- 472-1051
- 472-2184
- 472-2185
- 472-2195
- 472-2194
- 472-2191
- 472-2192
- 472-2250
- 472-2240
- 471-0231
- LIEBFKVSL5410
- 471-0232
- LIEBFKVSL2613
- 471-0234
- LIEBFKVSL3613
- LIEBFKVSL4112
- LIEBFKVSL5412
- LIEBFKVSL5413
- LIEBFK2640
- LIEBFK3640
- Temperatur:
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...12 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 2...12 °C
- 2...12 °C
- 2...12 °C
- 2...12 °C
- 2...12 °C
- 2...12 °C
- 2...12 °C
- 2...12 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 1...15 °C
- 2...10 °C
- 2...10 °C
- Luftcirkulation:
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ventileret
- ikke ventileret
- ikke ventileret
- ikke ventileret
- ikke ventileret
- ikke ventileret
- ikke ventileret
- ventileret
- ventileret
- Display:
- digital
- digital
- -
- Alarm:
- Audible and visual door and temperature alarm
- Audible and visual door and temperature alarm
- Audible and visual door and temperature alarm
- lydalarm og visuel alarm for dør og temperatur
- -
- -
- lydalarm og visuel alarm for dør og temperatur
- -
- -
- -
- -
- Audible and visual door and temperature alarm
- Audible and visual door and temperature alarm
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- Audible and visual door and temperature alarm
- -
- -
- -
- Hylder/skuffer:
- 3 trådnethylder
- 3 shelves
- 3 shelves
- 3 trådnethylder
- 4 shelves
- 5 trådnethylder
- 5 trådnethylder
- 4 shelves
- 2 trådnethylder
- 2 shelves
- 3 trådnethylder
- 3 shelves
- 3 shelves
- 3 trådnethylder
- 4 trådnethylder
- 5 trådnethylder
- 3 shelves
- 4 trådnethylder
- 5 trådnethylder
- 5 trådnethylder
- 3 trådnethylder
- 4 trådnethylder
- 5 trådnethylder
- 5 trådnethylder
- 3 trådnethylder
- 4 trådnethylder
- 5 shelves
- 6 shelves
- 5 shelves
- 5 shelves
- 6 shelves
- 6 shelves
- 6 shelves
- 6 shelves
- 6 shelves
- 4 shelves
- 5 shelves
- Dørtype:
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 solid swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 solid swing door
- 1 solid swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 glass swing door
- 1 solid swing door
- 1 solid swing door
- Dørlås:
- Yes
- Yes
- Yes
- Yes
- Yes
- ja
- Materiale:
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Rustfrit stål; Indvendigt: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Rustfrit stål; Indvendigt: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål, hvid; Interiør: ABS
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål; Interiør: Plastik
- Udvendigt: Stål; Interiør: Plastik
- B×D×H ud.:
- 600×615×830 mm
- 600×600×850 mm
- 600×610×1250 mm
- 600×610×1250 mm
- 600×650×1860 mm
- 750×730×1640 mm
- 730×750×1640 mm
- 430×500×610 mm
- 425×450×612 mm
- 600×615×830 mm
- 600×615×830 mm
- 600×615×830 mm
- 600×610×1250 mm
- 600×610×1640 mm
- 750×730×1640 mm
- 600×600×1220 mm
- 600×600×1590 mm
- 752×710×1516 mm
- 600×600×850 mm
- 600×600×1220 mm
- 600×600×1590 mm
- 752×710×1516 mm
- 600×600×855 mm
- 600×600×1210 mm
- 730×750×1640 mm
- 600×610×1250 mm
- Vægt:
- 35 kg
- 35 kg
- 35 kg
- 53 kg
- 49 kg
- 77 kg
- 74 kg
- 77 kg
- 34 kg
- 31 kg
- 35 kg
- 41 kg
- 42 kg
- 53 kg
- 56 kg
- 74 kg
- 45 kg
- 42 kg
- 55 kg
- 68 kg
- 45 kg
- 58 kg
- 55 kg
- 86 kg
- 50 kg
- 60 kg
- 51 kg
- 78 kg
- 67 kg
- 64 kg
- 82 kg
- 79 kg
- 86 kg
- 94 kg
- 99 kg
- 44 kg
- 53 kg
Specifikationer
Product Family Options
Køleskabe, med alm. dør, FKv og FKUv
Produktinformation
- KapacitetTemperaturB×D×H udv.TilgængelighedPrice
- 471-0471Professionelt køleskab, Premium, hvidt, FKUv 1610Professionelt køleskab, Premium, hvidt, FKUv 1610141 l1...15 °C600×615×830 mmLog ind for tilgængelighed1 * 1 Items9.099,00 krSpecifikationer:
More
Specifications
Cat. No.471-0471ModelFKUv 1610Art.nr471-0471Temperatur1...15 °CLuftcirkulationventileretDisplayAlarmAudible and visual door and temperature alarmHylder/skuffer3 trådnethylderDørtype1 solid swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×615×830 mmVægt35 kg - LIEBFKUV1660Professional refrigerator, Premium, stainless steel, FKUv 1660Professional refrigerator, Premium, stainless steel, FKUv 1660141 l1...15 °C600×615×830 mmLog ind for tilgængelighed1 * 1 Items9.699,00 krSpecifikationer:
More
Specifications
Cat. No.LIEBFKUV1660ModelFKUv 1660Art.nrLIEBFKUV1660Temperatur1...15 °CLuftcirkulationDisplayAlarmAudible and visual door and temperature alarmHylder/skuffer3 shelvesDørtype1 solid swing doorDørlåsMaterialeUdvendigt: Rustfrit stål; Indvendigt: ABSB×D×H ud.Vægt35 kg - 471-1064Professional under bench refrigerator, Premium, white, FKv 1800Professional under bench refrigerator, Premium, white, FKv 1800180 l1...15 °C600×600×850 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-1064ModelFKv 1800Art.nr471-1064Temperatur1...15 °CLuftcirkulationventileretDisplayAlarmAudible and visual door and temperature alarmHylder/skuffer3 shelvesDørtype1 solid swing doorDørlåsYesMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×600×850 mmVægt35 kg - 471-0225Professionelt køleskab, Premium, hvidt, FKv 2610Professionelt køleskab, Premium, hvidt, FKv 2610236 l1...15 °C600×610×1250 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-0225ModelFKv 2610Art.nr471-0225Temperatur1...15 °CLuftcirkulationventileretDisplaydigitalAlarmlydalarm og visuel alarm for dør og temperaturHylder/skuffer3 trådnethylderDørtype1 solid swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×610×1250 mmVægt53 kg - 471-1034Professional refrigerator, Premium, white, FKv 2640Professional refrigerator, Premium, white, FKv 2640240 l1...15 °C600×610×1250 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-1034ModelFKv 2640Art.nr471-1034Temperatur1...15 °CLuftcirkulationventileretDisplayAlarm-Hylder/skuffer4 shelvesDørtype1 solid swing doorDørlåsYesMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×610×1250 mmVægt49 kg - 472-0008Professionelt køleskab, Premium, hvidt, FKv 4310Professionelt køleskab, Premium, hvidt, FKv 4310421 l1...15 °C600×650×1860 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.472-0008ModelFKv 4310Art.nr472-0008Temperatur1...15 °CLuftcirkulationDisplayAlarm-Hylder/skuffer5 trådnethylderDørtype1 solid swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×650×1860 mmVægt77 kg - 471-0410Professionelt køleskab, Premium, hvidt, FKv 5410Professionelt køleskab, Premium, hvidt, FKv 5410544 l1...15 °C750×730×1640 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-0410ModelFKv 5410Art.nr471-0410Temperatur1...15 °CLuftcirkulationventileretDisplayAlarmlydalarm og visuel alarm for dør og temperaturHylder/skuffer5 trådnethylderDørtype1 solid swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.750×730×1640 mmVægt74 kg - 471-1030Professional refrigerator, Premium, white, FKv 5440Professional refrigerator, Premium, white, FKv 5440544 l1...15 °C730×750×1640 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-1030ModelFKv 5440Art.nr471-1030Temperatur1...15 °CLuftcirkulationventileretDisplayAlarm-Hylder/skuffer4 shelvesDørtype1 solid swing doorDørlåsYesMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.730×750×1640 mmVægt77 kg
Køleskabe, med glasdør, FKv og FKUv
Produktinformation
- KapacitetTemperaturB×D×H udv.TilgængelighedPrice
- 471-0230Professionelt køleskab, underskabsmodel, Premium, gråt, FKv 502Professionelt køleskab, underskabsmodel, Premium, gråt, FKv 50245 l1...15 °C430×500×610 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-0230ModelFKv 502Art.nr471-0230Temperatur1...15 °CLuftcirkulationDisplayAlarm-Hylder/skuffer2 trådnethylderDørtype1 glass swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.430×500×610 mmVægt34 kg - 471-1036Professional under bench, refrigerator, grey, FKv 503Professional under bench, refrigerator, grey, FKv 50345 l1...12 °C425×450×612 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-1036ModelFKv 503Art.nr471-1036Temperatur1...12 °CLuftcirkulationventileretDisplayAlarm-Hylder/skuffer2 shelvesDørtype1 glass swing doorDørlåsYesMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.425×450×612 mmVægt31 kg - 471-0693Professionelt køleskab, Premium, hvidt, FKUv 1612Professionelt køleskab, Premium, hvidt, FKUv 1612141 l1...15 °C600×615×830 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-0693ModelFKUv 1612Art.nr471-0693Temperatur1...15 °CLuftcirkulationventileretDisplayAlarm-Hylder/skuffer3 trådnethylderDørtype1 glass swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×615×830 mmVægt35 kg - 471-1062Professional under bench refrigerator, Premium, white, FKUv 1613Professional under bench refrigerator, Premium, white, FKUv 1613141 l1...15 °C600×615×830 mmLog ind for tilgængelighed1 * 1 Items9.799,00 krSpecifikationer:
More
Specifications
Cat. No.471-1062ModelFKUv 1613Art.nr471-1062Temperatur1...15 °CLuftcirkulationventileretDisplayAlarmAudible and visual door and temperature alarmHylder/skuffer3 shelvesDørtype1 glass swing doorDørlåsYesMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×615×830 mmVægt41 kg - 471-1180Professional under bench refrigerator, grey, FKUv 1663Professional under bench refrigerator, grey, FKUv 1663141 l1...15 °C600×615×830 mmLog ind for tilgængelighed1 * 1 Items11.649,00 krSpecifikationer:
More
Specifications
Cat. No.471-1180ModelFKUv 1663Art.nr471-1180Temperatur1...15 °CLuftcirkulationDisplayAlarmAudible and visual door and temperature alarmHylder/skuffer3 shelvesDørtype1 glass swing doorDørlåsMaterialeUdvendigt: Rustfrit stål; Indvendigt: ABSB×D×H ud.600×615×830 mmVægt42 kg - 471-1033Professionelt køleskab, Premium, hvidt, FKv 2643Professionelt køleskab, Premium, hvidt, FKv 2643236 l1...15 °C600×610×1250 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-1033ModelFKv 2643Art.nr471-1033Temperatur1...15 °CLuftcirkulationventileretDisplayAlarm-Hylder/skuffer3 trådnethylderDørtype1 glass swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×610×1250 mmVægt53 kg - 471-1031Professionelt køleskab, Premium, hvidt, FKv 3643Professionelt køleskab, Premium, hvidt, FKv 3643335 l1...15 °C600×610×1640 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-1031ModelFKv 3643Art.nr471-1031Temperatur1...15 °CLuftcirkulationventileretDisplayAlarm-Hylder/skuffer4 trådnethylderDørtype1 glass swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.600×610×1640 mmVægt56 kg - 471-1029Professionelt køleskab, Premium, hvidt, FKv 5443Professionelt køleskab, Premium, hvidt, FKv 5443544 l1...15 °C750×730×1640 mmDiscontinued-Specifikationer:
More
Specifications
Cat. No.471-1029ModelFKv 5443Art.nr471-1029Temperatur1...15 °CLuftcirkulationventileretDisplayAlarm-Hylder/skuffer5 trådnethylderDørtype1 glass swing doorDørlåsMaterialeUdvendigt: Stål, hvid; Interiør: ABSB×D×H ud.750×730×1640 mmVægt74 kg
Services
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice
- SERW1100.7000.01Installationskvalificering og driftskvalificering af køleskab on-site. Kvalifikationsprotokoller er ikke inkluderet. Rejseudgifter er inkluderet (ekskl. udgifter til flyrejser).Log ind for tilgængelighed1 * 1 Each - 42.950,00 kr
- SERW1100.0100.01Installation af køleskab on-site. Enheden skal være på anvendelsesstedet. Funktionstest og kort introduktion af brugernes korte funktioner. Rejseudgifter inkluderet (ekskl. udgifter til flyrejser).Log ind for tilgængelighed1 * 1 Each - 12.280,00 kr
- SERW1100.1101.01Forebyggende vedligeholdelse, driftsprøvning og kalibrering af køleskabet on-site. 1 Temperatur (1 probe) Kalibrering . Kalibreringscertifikat medfølger. Reservedele og rejseudgifter ikke inkluderet.Log ind for tilgængelighed1 * 1 Each - 3.090,00 kr
- SERW1100.0900.01Forebyggende vedligeholdelse og driftstest af køleskabet on-site. Reservedele og rejseudgifter er ikke inkluderet.Log ind for tilgængelighed1 * 1 Each - 2.060,00 kr