Orders LinkContactUsLinkComponent
Molecular model: organic set
Molecular model: organic set
Molecular organic model, 50
Katalog # 761-0111
Marketsource
Leverandør:   SPIRING ENTERPRISES
Molecular model: organic set
Katalog # 761-0111
Leverandør:  SPIRING ENTERPRISES
Marketsource
Molecular model: organic set
Molecular model: organic set
Molecular organic model, 50
Price
152,55 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
More detailed set for students at a more advanced level.
Atoms: 20 hydrogen (white), 12 carbon (black), 6 oxygen (red), 4 halogen (green), 4 nitrogen (blue), 2 sulphur (yellow), 1 phosphorus (purple), 1 metal (grey).
Bonds: 26 medium grey, 10 long flexible grey, 26 short white; one link remover tool.
Emballering: In a plastic box with instructions for use.
Specifikationer
  • Beskrivelse:
    Molecular organic model
  • Antal atomer:
    50