Reading magnifiers
Leverandør: ESCHENBACH OPTIK
Gå til:
Om denne vare
This magnifier provides biconvex PXM® lightweight lens for an ergonomic use and optimised performances.
- Additional plano-convex lens in the handle for 5× magnification
- Eyelet in the handle, round-the-neck carry cord supplied
- Antistatic coating
2 tilgængelige muligheder nedenfor
Product Details & Documents
This magnifier provides biconvex PXM® lightweight lens for an ergonomic use and optimised performances.
- Additional plano-convex lens in the handle for 5× magnification
- Eyelet in the handle, round-the-neck carry cord supplied
- Antistatic coating