Homogenisator, T 10 basic ULTRA-TURRAX®
Om denne vare
Compact, hand-held tissue homogenizer for volumes from 0,5 to 100 ml with a quick-connect coupling for easy interchangeability of dispersing tools.
Quick-connect coupling for changing dispersing elements easily
- Ideal for manual operation due to its light weight and ergonomic design
- Extremely mobile due to direct line power (no transformer required)
- Stainless steel dispersing elements can be cleaned quickly and easily as they can be dismounted without tools
- Plastic disposable dispersing elements are available in two sizes, particularly suitable for PCR analysis
- High performance drive ensures immense speed stability
- Quiet operation
Small dispersing instrument for volumes from 0,5 to 100 ml (H₂O). It offers a wide speed range from 8000 to 30,000 min⁻¹ that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
Certifications: EN 61010-1:2010 + A1:2019 + A1:2019/AC:2019
EN 61010-2-051:2015
EN 61326-1:2013
EN 60529:1991 + A1:2000 + A2:2013
EN ISO 12100:2010
2014/35/EU Low Voltage Directive
2006/42/EG Machinery Directive
2014/30/EU EMC Directive
2011/65/EU RoHS Directive
Accessories information: The drive unit always requires a dispersing tool, please choose from the compatible tools.
Bestillingsoplysninger: The dispersing tool is not included in the scope of delivery. Packages include Stands and Dispersing tools.
Leveringsinformation: Supplied with T 10 basic ULTRA-TURRAX® and R 200 clamp.
1 tilgængelige muligheder nedenfor
Product Details & Documents
Compact, hand-held tissue homogenizer for volumes from 0,5 to 100 ml with a quick-connect coupling for easy interchangeability of dispersing tools.
Quick-connect coupling for changing dispersing elements easily
- Ideal for manual operation due to its light weight and ergonomic design
- Extremely mobile due to direct line power (no transformer required)
- Stainless steel dispersing elements can be cleaned quickly and easily as they can be dismounted without tools
- Plastic disposable dispersing elements are available in two sizes, particularly suitable for PCR analysis
- High performance drive ensures immense speed stability
- Quiet operation
Small dispersing instrument for volumes from 0,5 to 100 ml (H₂O). It offers a wide speed range from 8000 to 30,000 min⁻¹ that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
Certifications: EN 61010-1:2010 + A1:2019 + A1:2019/AC:2019
EN 61010-2-051:2015
EN 61326-1:2013
EN 60529:1991 + A1:2000 + A2:2013
EN ISO 12100:2010
2014/35/EU Low Voltage Directive
2006/42/EG Machinery Directive
2014/30/EU EMC Directive
2011/65/EU RoHS Directive
Accessories information: The drive unit always requires a dispersing tool, please choose from the compatible tools.
Bestillingsoplysninger: The dispersing tool is not included in the scope of delivery. Packages include Stands and Dispersing tools.
Leveringsinformation: Supplied with T 10 basic ULTRA-TURRAX® and R 200 clamp.
Specifications for Product Family
Specifikationer
- Catalog No:
- 431-0179
- 431-0188
- 431-0189
- Model:
- T 10 basic
- T 10 basic
- T 10 basic
- Volumen H₂O:
- 0,5 - 100 ml
- 0,5 - 100 ml
- 0,5 - 100 ml
- Maks. viskositet:
- 5000 mPas
- 5000 mPas
- 5000 mPas
- Hastighedsområde:
- 8000 - 30000 min⁻¹
- 8000 - 30000 min⁻¹
- 8000 - 30000 min⁻¹
- Hastighedsvisning:
- Scale
- Scale
- Scale
- Effekt, motorindgang/-udgang:
- 125/75 W
- 125/75 W
- 125/75 W
- Vægt:
- 0,5 kg
- 0,5 kg
- 0,5 kg
- B×D×H:
- 56×66×178 mm
- 56×66×178 mm
- 56×66×178 mm
Specifikationer
Product Family Options
T 10 basic ULTRA-TURRAX® Dispersing drive unit
Produktinformation
- TilgængelighedPrice
- 431-0179T 10 basic ULTRA-TURRAX® homogeniser, EU-plugSpecifikationer:
More
Specifications
Cat. No.431-0179ModelT 10 basicVolumen H₂O0,5 - 100 mlMaks. viskositet5000 mPasHastighedsområde8000 - 30000 min⁻¹HastighedsvisningScaleEffekt, motorindgang/-udgang125/75 WVægt0,5 kgB×D×H56×66×178 mm - 431-0188T 10 basic ULTRA-TURRAX® homogeniser, UK-plugT 10 basic ULTRA-TURRAX® homogeniser, UK-plugDiscontinued-Specifikationer:
More
Specifications
Cat. No.431-0188ModelT 10 basicVolumen H₂O0,5 - 100 mlMaks. viskositet5000 mPasHastighedsområde8000 - 30000 min⁻¹HastighedsvisningScaleEffekt, motorindgang/-udgang125/75 WVægt0,5 kgB×D×H56×66×178 mm - 431-0189T 10 basic ULTRA-TURRAX® homogeniser, CH-plugT 10 basic ULTRA-TURRAX® homogeniser, CH-plugDiscontinued-Specifikationer:
More
Specifications
Cat. No.431-0189ModelT 10 basicVolumen H₂O0,5 - 100 mlMaks. viskositet5000 mPasHastighedsområde8000 - 30000 min⁻¹HastighedsvisningScaleEffekt, motorindgang/-udgang125/75 WVægt0,5 kgB×D×H56×66×178 mm
Dispersing tools
Produktinformation
- Katalog #HastighedVolumenBeskrivelseStator/rotor ØLængdeTilTilgængelighedPrice
- 7,5 m/s1 - 40 mlS10D-7G-KS-1107 / 4,8 mm110 mmT 10 basicLog ind for tilgængelighed1 * 24 Items5.010,00 kr
Stands
Produktinformation
- Katalog #TilBeskrivelseTilgængelighedPrice
- T 10 basic/VDI 12Pladestativ, 200×355 mm, stang 10 mm Ø, højde 370 mmLog ind for tilgængelighed1 * 1 Items1.670,00 kr
Accessories
Produktinformation
- Katalog #TilTilgængelighedPrice
- R200 clamp for fastening the T 10 basic to the standR200 clamp for fastening the T 10 basic to the stand442-0313T 10 basicLog ind for tilgængelighed1 * 1 Items - 364,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Dobbeltmuffe
Produktinformation
- Katalog #TilgængelighedPrice
- Beaker cap, BC 1000, silicone, for 600 and 1000 ml beakersBeaker cap, BC 1000, silicone, for 600 and 1000 ml beakersIKAA20003417Log ind for tilgængelighed1 * 1 Items - 180,00 kr