Electrical sandbaths, LHG
Om denne vare
Aluminium alloy plate, stainless steel housing with variable temperature control.
- Robust construction for continuous performance
- Even temperature is guaranteed over the entire heating surface
Sealed stainless steel frame with a usable height of 50 mm. The sand lays directly in contact with the hotplate. Four adjustable feet ensure extra stability.
Caution: With thermostatic regulator.
14 tilgængelige muligheder nedenfor
Product Details & Documents
Aluminium alloy plate, stainless steel housing with variable temperature control.
- Robust construction for continuous performance
- Even temperature is guaranteed over the entire heating surface
Sealed stainless steel frame with a usable height of 50 mm. The sand lays directly in contact with the hotplate. Four adjustable feet ensure extra stability.
Caution: With thermostatic regulator.
Specifications for Product Family
Specifikationer
- Catalog No:
- 453-0039
- 453-0040
- 453-0089
- GESTST71
- 453-2211
- 453-2214
- GESTST81
- 453-2212
- GESTST83
- GESTST91
- 453-2213
- GESTST92-400
- GESTST93-230
- GESTST93-400
- Model:
- SD 7
- SD 8
- SD 9-230
- ST 71*
- ST72*
- ST 73*
- ST 81*
- ST 82*
- ST 83*
- ST 91*
- ST 92-230*
- ST 92-400*
- ST 93-230*
- ST 93-400*
- Temperaturinterval:
- Max. 370 °C
- Max. 370 °C
- Max. 370 °C
- 30…110 °C
- 50…300 °C
- 130…370 °C
- 30…110 °C
- 50…300 °C
- 50…300 °C
- 50…300 °C
- 50…300 °C
- 50…300 °C
- 50…300 °C
- 50…300 °C
- Effekt:
- 2200 W
- 2850 W
- 4000 W
- 1150 W
- 2200 W
- 2200 W
- 1800 W
- 2850 W
- 2850 W
- 2000 W
- 4000 W
- 4000 W
- 4000 W
- 4000 W
- Bad B×D×H:
- 360×360×50 mm
- 440×590×50 mm
- 440×590×50 mm
- 360×360×50 mm
- 360×360×50 mm
- 360×360×50 mm
- 360×510×50 mm
- 360×510×50 mm
- 360×510×50 mm
- 440×590×50 mm
- 440×590×50 mm
- 440×590×50 mm
- 440×590×50 mm
- 440×590×50 mm
- B×D×H:
- 360×360×220 mm
- 440×514×220 mm
- 442×592×220 mm
- 360×360×220 mm
- 360×360×220 mm
- 360×360×220 mm
- 364×514×220 mm
- 364×514×220 mm
- 364×514×220 mm
- 442×592×220 mm
- 442×592×220 mm
- 442×592×220 mm
- 442×592×220 mm
- 442×592×220 mm
- Vægt:
- 13 kg
- 21 kg
- 27 kg
- 14 kg
- 14 kg
- 14 kg
- 21 kg
- 21 kg
- 21 kg
- 28 kg
- 28 kg
- 28 kg
- 28 kg
- 28 kg
Specifikationer
Product Family Options
Produktinformation
- Opvarmnings kapacitetPladestørrelseModelTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.453-0039ModelSD 7TemperaturintervalMax. 370 °CEffekt2200 WBad B×D×H360×360×50 mmB×D×H360×360×220 mmVægt13 kg - Specifikationer:
More
Specifications
Cat. No.453-0040ModelSD 8TemperaturintervalMax. 370 °CEffekt2850 WBad B×D×H440×590×50 mmB×D×H440×514×220 mmVægt21 kg - Specifikationer:
More
Specifications
Cat. No.453-0089ModelSD 9-230TemperaturintervalMax. 370 °CEffekt4000 WBad B×D×H440×590×50 mmB×D×H442×592×220 mmVægt27 kg - Specifikationer:
More
Specifications
Cat. No.GESTST71ModelST 71*Temperaturinterval30…110 °CEffekt1150 WBad B×D×H360×360×50 mmB×D×H360×360×220 mmVægt14 kg - Specifikationer:
More
Specifications
Cat. No.453-2211ModelST72*Temperaturinterval50…300 °CEffekt2200 WBad B×D×H360×360×50 mmB×D×H360×360×220 mmVægt14 kg - Specifikationer:
More
Specifications
Cat. No.453-2214ModelST 73*Temperaturinterval130…370 °CEffekt2200 WBad B×D×H360×360×50 mmB×D×H360×360×220 mmVægt14 kg - Specifikationer:
More
Specifications
Cat. No.GESTST81ModelST 81*Temperaturinterval30…110 °CEffekt1800 WBad B×D×H360×510×50 mmB×D×H364×514×220 mmVægt21 kg - Specifikationer:
More
Specifications
Cat. No.453-2212ModelST 82*Temperaturinterval50…300 °CEffekt2850 WBad B×D×H360×510×50 mmB×D×H364×514×220 mmVægt21 kg - Specifikationer:
More
Specifications
Cat. No.GESTST83ModelST 83*Temperaturinterval50…300 °CEffekt2850 WBad B×D×H360×510×50 mmB×D×H364×514×220 mmVægt21 kg - Specifikationer:
More
Specifications
Cat. No.GESTST91ModelST 91*Temperaturinterval50…300 °CEffekt2000 WBad B×D×H440×590×50 mmB×D×H442×592×220 mmVægt28 kg - Specifikationer:
More
Specifications
Cat. No.453-2213ModelST 92-230*Temperaturinterval50…300 °CEffekt4000 WBad B×D×H440×590×50 mmB×D×H442×592×220 mmVægt28 kg - Specifikationer:
More
Specifications
Cat. No.GESTST92-400ModelST 92-400*Temperaturinterval50…300 °CEffekt4000 WBad B×D×H440×590×50 mmB×D×H442×592×220 mmVægt28 kg - Specifikationer:
More
Specifications
Cat. No.GESTST93-230ModelST 93-230*Temperaturinterval50…300 °CEffekt4000 WBad B×D×H440×590×50 mmB×D×H442×592×220 mmVægt28 kg - Specifikationer:
More
Specifications
Cat. No.GESTST93-400ModelST 93-400*Temperaturinterval50…300 °CEffekt4000 WBad B×D×H440×590×50 mmB×D×H442×592×220 mmVægt28 kg
Accessories
Produktinformation
- Katalog #Opvarmnings kapacitetPladestørrelseTilgængelighedPrice
- Sand bath filling, fire-dried quartz sand, grain size 0,1 - 0,4 mmSand bath filling, fire-dried quartz sand, grain size 0,1 - 0,4 mm453-22324000 W440×590 mmLog ind for tilgængelighed1 * 4 kg - 123,00 kr