VWR®, Software til validering af analytiske metoder, Valideringsmanager
HPLC1.18988.SMA1
- Beskrivelse:SMA (Software Maintenance Agreement - Softwarevedligeholdelsesaftale) til Validation Manager, enkeltbrugerlicens
- Environmentally Preferable:
- Pakn:1
Validation Manager-softwaren kontrollerer, om din analysemetode er egnet til den tiltænkte anvendelse, og producerer automatisk den valideringsrapport, du skal bruge. Den virtuelle Validation Manager kan spare dig for dages eller endog ugers valideringsarbejde med enhver analyseteknik.
- Based on the international guidelines for method validation
- Versatile configuration for all analytical techniques
- Ease of operation and easy data entry
- Automatic calculation and evaluation of the method characteristics
- Automatic report compilation
- Validated and fully FDA compliant
For further information on Validation Manager software, please contact the specialists at Avantor or email [email protected].