Passive gas detectors, Dositubes
Om denne vare
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
- Compact, lightweight, and versatile (can be used anywhere)
- Easy direct measuring and reading
- No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
22 tilgængelige muligheder nedenfor
Product Details & Documents
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
- Compact, lightweight, and versatile (can be used anywhere)
- Easy direct measuring and reading
- No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
Specifications for Product Family
Specifikationer
- Catalog No:
- 151-0089
- 151-0084
- 151-0090
- 151-0093
- 151-0078
- 151-0076
- 151-0080
- 151-0079
- 151-0083
- 151-0082
- 151-0075
- 151-0074
- 151-0081
- 151-0086
- 151-0094
- 151-0088
- 151-0073
- 151-0091
- 151-0062
- 151-0077
- 151-0072
- 151-0087
- Måletid:
- 1 - 5 h
- 1 - 10 h
- 1 - 10 h
- 1 - 10 h
- 1 -10 h
- 1 - 8 h
- 1 - 8 h
- 1 - 8 h
- 0,5 - 10 h
- 0,5 - 24 h
- 1 - 8 h
- 1 - 8 h
- 0,5 - 48 h
- 0,5 - 10 h
- 1 - 48 h
- 1 - 10 h
- 1 - 10 h
- 0,5 - 24 h
- 1 - 10 h
- 1 - 10 h
- 1 - 10 h
- 1 - 10 h
Specifikationer
Product Family Options
Produktinformation
- ParameterOmrådeTilgængelighedPrice
- 151-0089Svovldioxid (5DH)Specifikationer:
More
Specifications
Cat. No.151-0089Måletid1 - 5 h - 151-0084Hydrogenperoxid (32D)Specifikationer:
More
Specifications
Cat. No.151-0084Måletid1 - 10 h - 151-0090Eddikesyre (81D)Specifikationer:
More
Specifications
Cat. No.151-0090Måletid1 - 10 h - 151-0093Nitrogendioxid (9D)Specifikationer:
More
Specifications
Cat. No.151-0093Måletid1 - 10 h - 151-0078Methylethylketon (152D)Methylethylketon (152D)Methyl ethyl ketone2 - 600 ppmLog ind for tilgængelighed1 * 10 Items486,00 krSpecifikationer:
More
Specifications
Cat. No.151-0078Måletid1 -10 h - 151-0076Hydrogenchlorid (14D)Specifikationer:
More
Specifications
Cat. No.151-0076Måletid1 - 8 h - 151-0080Hydrogenfluorid (17D)Specifikationer:
More
Specifications
Cat. No.151-0080Måletid1 - 8 h - 151-0079Butadien (174D)Specifikationer:
More
Specifications
Cat. No.151-0079Måletid1 - 8 h - 151-0083Kuldioxid (5D)Specifikationer:
More
Specifications
Cat. No.151-0083Måletid0,5 - 10 h - 151-0082Kulmonoxid (1DLSpecifikationer:
More
Specifications
Cat. No.151-0082Måletid0,5 - 24 h - 151-0075Perchlorethylen (133D)Specifikationer:
More
Specifications
Cat. No.151-0075Måletid1 - 8 h - 151-0074Trichlorethylen (132D)Specifikationer:
More
Specifications
Cat. No.151-0074Måletid1 - 8 h - 151-0081Kulmonoxid (1D)Specifikationer:
More
Specifications
Cat. No.151-0081Måletid0,5 - 48 h - 151-0086Ammoniak (3D)Specifikationer:
More
Specifications
Cat. No.151-0086Måletid0,5 - 10 h - 151-0094Hydrogensulfid (4D)Specifikationer:
More
Specifications
Cat. No.151-0094Måletid1 - 48 h - 151-0088Svovldioxid (5D)Specifikationer:
More
Specifications
Cat. No.151-0088Måletid1 - 10 h - 151-0073HCN (12D)Specifikationer:
More
Specifications
Cat. No.151-0073Måletid1 - 10 h - 151-0091Chlor (8D)Specifikationer:
More
Specifications
Cat. No.151-0091Måletid0,5 - 24 h - 151-0062Formaldehyd (91D)Specifikationer:
More
Specifications
Cat. No.151-0062Måletid1 - 10 h - 151-0077Acetone (151D)Specifikationer:
More
Specifications
Cat. No.151-0077Måletid1 - 10 h - 151-0072Toluen (122D)Specifikationer:
More
Specifications
Cat. No.151-0072Måletid1 - 10 h - 151-0087Ammoniak (3DL)Specifikationer:
More
Specifications
Cat. No.151-0087Måletid1 - 10 h
Accessories
Produktinformation
- Katalog #ParameterOmrådeTilgængelighedPrice
- Eddikesyre (81D)Eddikesyre (81D)151-0090Acetic acid0,5 - 100 ppmLog ind for tilgængelighed1 * 10 Items - 486,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Holder til Dositube