Airflow detection powder, Draft Checker™, SP Bel-Art
Catalog # 215-6525
Supplier: Bel-Art Products, a Part of SP
Specifications
- Beskrivelse:Draft Checker™ 1,5 g
- Pakn:1
Specifications
About this item
An amorphous ultra-fine silica powder in a handy squeezy bottle.
- Suspended powder permits visual estimation of airflow
- Can be used for detecting leaks around windows, doors and ducts
Safety Note: Silica is inert and non-flammable but appropriate precautions should be taken to prevent ingestion or inhalation of the powder.