pH and redox combination electrodes with temperature sensor, BlueLine
662-6139
- Tilslutning:DIN + bananstik
- Ø×L:12×120 mm
- Model:24 pH, with NTC 30 kΩ temp. sensor
- Beskrivelse:Standardelektrode 24 pH, NTC30-sensor
- Materiale:Plast
- Seq:03
- Elektrolyt:Gel
- Pakn:1
- Temperatur:−5...+80 °C
- pH:0 - 14
The BlueLine family comprises three ranges of combination electrodes: Robust electrodes with gel electrolyte and plastic shaft for general use, liquid-electrolyte sensors for more critical measurements and special sensors.
- Special electrodes for redox measurements, surface measurements and food applications
- Gel electrolyte requires low maintenance and is not refillable
- ID sensors are equipped with a transponder, carrying the information of its type and serial number as well as the date and result of the last calibration
- IDS sensors automatically identify themselves with their serial numbers and type designation and transmit their calibration data when connected to a measuring device
pH range: 0 - 14