Kitchen rolls, Tork Premium
2 ply, 64 sheets, white, 15 m×230 mm, 14×15 m ruller
Katalog # 115-0330
Leverandør: Essity
Gå til:
Product Details & Documents
High quality kitchen roll for wiping and cleaning. A flexible alternative in many kitchen environments.
- Excellent absorption
- Can be disposed of with household waste
Certifications: ISO 9001:2000, ISO 14001 and EMAS.
Specifikationer
- Pakket:14×15 m ruller
- Beskrivelse:2 ply, 64 sheets, white
- L×B:15 m×230 mm