Gulvservietruller, Multizell®
380×240 mm, 10 ruller af 100
Katalog # 115-2736
Leverandør: Zellstoff-Vertriebs-GmbH & Co. KG
Gå til:
Product Details & Documents
These absorbent, 3-ply, low-lint wipes are made from unbleached and completely natural 100% recycled material. Also suitable for gas and repair stations.
- Perforated
- Highly absorbent
Specifikationer
- Pakket:10 ruller af 100
- L×B:380×240 mm