Viscosity cups with nozzle
Leverandør: Industrial Physics
Gå til:
Om denne vare
Anodised aluminium and stainless steel viscometer cups with nozzle for break-thread flow time comparative measurements.
- Easy cleaning
- Engraved with a unique serial number
Typically used for the consistency of paintings, varnishes and similar products.
Certifications: DIN 53211
Ordering information: The stand should be ordered separately.
6 tilgængelige muligheder nedenfor
Product Details & Documents
Anodised aluminium and stainless steel viscometer cups with nozzle for break-thread flow time comparative measurements.
- Easy cleaning
- Engraved with a unique serial number
Typically used for the consistency of paintings, varnishes and similar products.
Certifications: DIN 53211
Ordering information: The stand should be ordered separately.
Documents
SIMEVF1999_TDS_EN
Datasheet
TQC Company Video
Videos
Product Family Options
Produktinformation
- BægeråbningTilgængelighedPrice
- SIMEVF2000Viscosity cup with nozzle, 100 ml
- SIMEVF2001Viscosity cup with nozzle, 100 ml
- 769-1110Viskositetsflowkop
- SIMEVF2015Viscosity cup with nozzle, 100 ml
- 769-1121Viscosity cup with nozzle, 100 ml
- 769-1125Viscosity cup with nozzle, 100 ml
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Trefod (260 mm)
Produktinformation
- Katalog #TilgængelighedPrice
- Ring stand fro flow cups
Produktinformation
- Katalog #TilgængelighedPrice
- Digital stopwatch, 0,01 s
Produktinformation
- Katalog #TilgængelighedPrice
- Temperature control jacket with tripod for DIN and ASTM cupsTemperature control jacket with tripod for DIN and ASTM cups769-1131Log ind for tilgængelighed1 * 1 Items - 4.290,00 kr