Orders LinkContactUsLinkComponent
Slanger, PTFE
Supplier:   Bohlender
Slanger, PTFE
Supplier:  Bohlender
Certificates
Slanger, PTFE
52 Options Available Below
Product Details & Documents
PTFE, hvid, semi-transparent.
  • Fremragende kemisk resistens
  • Meget god termisk resistens, autoklaverbar
  • Fri for ekstraherbare stoffer, fysiologisk neutral, biokompatibel
Denne slange kendetegnes ved sin slip-let-overflade og fremragende dielektriske egenskaber.
Temperaturresistent fra −200 °C til +260 °C
Product Family Options
Product Information
  • Rør int.Ø
    Vægtykkelse
    Rør udv.Ø
    Availability
    Price
  • 0,20 mm
    0,10 mm
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr43.50
    kr43.50
    1 * 1 m
       kr43.50
  • 0,20 mm
    0,70 mm
    1,60 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr90.40
    kr90.40
    1 * 1 m
       kr90.40
  • 0,30 mm
    0,15 mm
    0,60 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr24.90
    kr24.90
    1 * 1 m
       kr24.90
  • 0,30 mm
    0,65 mm
    1,60 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr88.90
    kr88.90
    1 * 1 m
       kr88.90
  • 0,40 mm
    0,25 mm
    0,90 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr33.40
    kr33.40
    1 * 1 m
       kr33.40
  • 0,50 mm
    0,25 mm
    1,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr34.80
    kr34.80
    1 * 1 m
       kr34.80
  • 0,50 mm
    0,55 mm
    1,60 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr48.90
    kr48.90
    1 * 1 m
       kr48.90
  • 0,80 mm
    0,40 mm
    1,58 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr41.80
    kr41.80
    1 * 1 m
       kr41.80
  • 1,00 mm
    0,50 mm
    2,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr47.70
    kr47.70
    1 * 1 m
       kr47.70
  • 1,00 mm
    0,30 mm
    1,60 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr30.50
    kr30.50
    1 * 1 m
       kr30.50
  • 1,00 mm
    1,00 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr105.00
    kr105.00
    1 * 1 m
       kr105.00
  • 1,20 mm
    0,40 mm
    1,80 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr37.10
    kr37.10
    1 * 1 m
       kr37.10
  • 1,40 mm
    0,40 mm
    2,20 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr60.20
    kr60.20
    1 * 1 m
       kr60.20
  • 1,50 mm
    0,30 mm
    2,10 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr44.40
    kr44.40
    1 * 1 m
       kr44.40
  • 1,50 mm
    0,50 mm
    2,50 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr66.80
    kr66.80
    1 * 1 m
       kr66.80
  • 1,50 mm
    1,00 mm
    3,50 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr132.00
    kr132.00
    1 * 1 m
       kr132.00
  • 1,50 mm
    0,75 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr117.00
    kr117.00
    1 * 1 m
       kr117.00
  • 1,60 mm
    0,40 mm
    2,40 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr62.20
    kr62.20
    1 * 1 m
       kr62.20
  • 1,60 mm
    0,80 mm
    3,20 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr93.10
    kr93.10
    1 * 1 m
       kr93.10
  • 1,90 mm
    0,30 mm
    2,50 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr55.30
    kr55.30
    1 * 1 m
       kr55.30
  • 10,0 mm
    1,00 mm
    12,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr439.00
    kr439.00
    1 * 1 m
       kr439.00
  • 10,0 mm
    2,00 mm
    14,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr1,090.00
    kr1,090.00
    1 * 1 m
       kr1,090.00
  • 12,0 mm
    1,00 mm
    14,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr538.00
    kr538.00
    1 * 1 m
       kr538.00
  • 12,0 mm
    2,00 mm
    16,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr1,450.00
    kr1,450.00
    1 * 1 m
       kr1,450.00
  • 13,0 mm
    1,50 mm
    16,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr1,120.00
    kr1,120.00
    1 * 1 m
       kr1,120.00
  • 14,0 mm
    1,00 mm
    16,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr703.00
    kr703.00
    1 * 1 m
       kr703.00
  • 16,0 mm
    1,00 mm
    18,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr870.00
    kr870.00
    1 * 1 m
       kr870.00
  • 18,0 mm
    1,00 mm
    20,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr1,080.00
    kr1,080.00
    1 * 1 m
       kr1,080.00
  • 2,00 mm
    0,50 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr77.60
    kr77.60
    1 * 1 m
       kr77.60
  • 2,00 mm
    1,00 mm
    4,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr145.00
    kr145.00
    1 * 1 m
       kr145.00
  • 2,40 mm
    0,40 mm
    3,20 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr68.40
    kr68.40
    1 * 1 m
       kr68.40
  • 20,0 mm
    1,00 mm
    22,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr1,080.00
    kr1,080.00
    1 * 1 m
       kr1,080.00
  • 3,00 mm
    0,50 mm
    4,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr91.90
    kr91.90
    1 * 1 m
       kr91.90
  • 3,00 mm
    1,00 mm
    5,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr196.00
    kr196.00
    1 * 1 m
       kr196.00
  • 3,00 mm
    1,50 mm
    6,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr333.00
    kr333.00
    1 * 1 m
       kr333.00
  • 3,17 mm
    1,58 mm
    6,35 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr305.00
    kr305.00
    1 * 1 m
       kr305.00
  • 4,00 mm
    0,50 mm
    5,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr120.00
    kr120.00
    1 * 1 m
       kr120.00
  • 4,00 mm
    1,00 mm
    6,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr204.00
    kr204.00
    1 * 1 m
       kr204.00
  • 4,35 mm
    1,00 mm
    6,35 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr227.00
    kr227.00
    1 * 1 m
       kr227.00
  • 4,78 mm
    0,79 mm
    6,35 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr247.00
    kr247.00
    1 * 1 Items
       kr247.00
  • 5,00 mm
    0,50 mm
    6,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr160.00
    kr160.00
    1 * 1 m
       kr160.00
  • 5,00 mm
    1,00 mm
    7,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr264.00
    kr264.00
    1 * 1 m
       kr264.00
  • 6,00 mm
    0,50 mm
    7,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr165.00
    kr165.00
    1 * 1 m
       kr165.00
  • 6,00 mm
    1,00 mm
    8,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr263.00
    kr263.00
    1 * 1 m
       kr263.00
  • 7,00 mm
    0,50 mm
    8,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr179.00
    kr179.00
    1 * 1 m
       kr179.00
  • 7,00 mm
    1,00 mm
    9,00 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr406.00
    kr406.00
    1 * 1 m
       kr406.00
  • 7,50 mm
    1,25 mm
    10,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr561.00
    kr561.00
    1 * 1 m
       kr561.00
  • 8,00 mm
    0,75 mm
    9,50 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr321.00
    kr321.00
    1 * 1 m
       kr321.00
  • 8,00 mm
    1,00 mm
    10,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr373.00
    kr373.00
    1 * 1 m
       kr373.00
  • 8,00 mm
    2,00 mm
    12,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr835.00
    kr835.00
    1 * 1 m
       kr835.00
  • 8,00 mm
    1,50 mm
    11,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr728.00
    kr728.00
    1 * 1 m
       kr728.00
  • 9,00 mm
    1,00 mm
    11,0 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - kr407.00
    kr407.00
    1 * 1 m
       kr407.00