Luftudtag
4 mm, 180 mm
201-2610
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Hul, modvirker stænk ved kogning.
- Ø:4 mm
- Længde:180 mm