Closures for test tubes and flasks
Leverandør: JAECE INDUSTRIES
Gå til:
Om denne vare
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.
7 tilgængelige muligheder nedenfor
Product Details & Documents
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.
Product Family Options
Identi-plug
Produktinformation
- TilgængelighedPrice
- JAECL800-AIdenti-plug foam plugs, Ø 6 - 13 mm
- JAECL800-BIdenti-plug foam plugs, Ø 14 - 20 mm
- 734-1262Identi-plug skumpropper, Ø 21 - 26 mm
- JAECL800-DIdenti-plug foam plugs, Ø 35 - 45 mm
Cotton cheesecloth
Produktinformation
- TilgængelighedPrice
- 734-1264Bomuldsostelærred, kvalitet 50
This product has been blocked by your organization. Please contact your purchasing department for more information
Bleached cotton balls
Produktinformation
- TilgængelighedPrice
- 734-1266Vatkugler, medium, ikke-sterile
- 734-1265Vatkugler, store, ikke-sterile
This product has been blocked by your organization. Please contact your purchasing department for more information