Mikrocentrifugerør, 1,5 ml
1,5 ml, ufarvet, With attached lid, Graduated, conical bottom, non sterile
Katalog # 211-2610
Leverandør: Brand
Gå til:
Product Details & Documents
PP, med inddeling (kun 211-2610 og 212-0198), autoklaverbar, med fastgjort låg (undtagen 211-2611).
- Can withstand centrifugation up to 20000×g (except 211-2611: Up to 6000×g)
- Flat cap can be easily pierced
- Frosted marking area (only 211-2610 and 212-0198)
- Amber coloured microcentrifuge tubes are ideal for light-sensitive reagents
IVD (except 211-2611): CE-marked according to IVD-Directive 98/79 EC
Certifications: CE-mærket (kun 211-2610 og 212-0198) i henhold til IVD-direktiv 98/79 EF
Documents
Brand Microcentrifuge tubes with lid closure
Produktlitteratur
BRND780525_TDS_EN
Datablad
Specifikationer
- Model:With attached lid
- Version:Graduated, conical bottom, non sterile
- Kapacitet:1,5 ml
- Farve:ufarvet