Centrifuge tubes
18 ml, 17×112 mm, uden cap
Katalog # 525-0036
Leverandør: MURRAY S
Gå til:
Product Details & Documents
Glass, non-graduated, round bottom, without rim, without cap.
Specifikationer
- Lukkestørrelse:uden cap
- Ø×L:17×112 mm
- Kapacitet:18 ml