Orders LinkContactUsLinkComponent
General purpose thermometers for flasks
Leverandør:   Amarell
General purpose thermometers for flasks
Leverandør:  Amarell
Certifikater
General purpose thermometers for flasks
General purpose thermometers for flasks
2 tilgængelige muligheder nedenfor
Product Details & Documents
These strengthened in-glass thermometers provides reinforced tips for stirring applications.
  • Fitted anti-roll plastic cap with cord loop
  • White enamelled back
  • No breakable glass ring
  • Bulb integral with stem
  • 620-0121 and 620-0122 calibrated at four and five points respectively
Kviksølv.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Product Family Options
Produktinformation
  • Område
    Inddeling
    Længde
    Påfyldning
    Neds.- dybde
    Ø
    Tilgængelighed
    Price
  • ME9010
    −10...+50 °C
    0,5 °C
    305 mm
    Kviksølv
    76 mm
    6,0 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • ME9008
    −10...+110 °C
    1,0 °C
    305 mm
    Kviksølv
    76 mm
    6,0 mm