Testpapir, Supelco®
Lead(II) acetate paper, 4,8 m roll
Leverandør: Merck
Gå til:
Product Details & Documents
Lead (II) acetate paper is used for the determination of sulphide and hydrogen sulphide.
Specifikationer
- Beskrivelse:Lead(II) acetate paper, 4,8 m roll
- Cat. No.:1.09511.0003
- M-Clarity quality level:MQ100