Orders LinkContactUsLinkComponent
Sterican® Single-Use Needles
Sterican® Single-Use Needles
Leverandør:   B. Braun
Sterican® Single-Use Needles
Leverandør:  B. Braun
Certifikater
Sterican® Single-Use Needles
Sterican® Single-Use Needles
2 tilgængelige muligheder nedenfor
Product Details & Documents
Stainless chromium-nickel steel needle with extremely smooth surface and light silicone coating, transparent Luer lock PP hub.
  • Colour coded hub, conforms to ISO 6009
  • Latex- and PVC-free
  • Tri-bevelled for minimal pain upon puncture (long bevel)
Certifications: ISO 7864, DIN 13097
Packaging: Individually, sterile packed. 100 needles (20 blocks of 5) per box.
Documents
Product Family Options
Sterile
Produktinformation
  • Måler
    Længde
    Ø
    Farvekode
    Tilgængelighed
    Price
  • BRAU4657527B
    Intravenous, for large volumes, intramuscular, for hydrous solutions (size 2), made in china, with Max.shelf life 60 months
    Intravenous, for large volumes, intramuscular, for hydrous solutions (size 2), made in china, with Max.shelf life 60 months
    21
    40 mm
    0,80 mm
    grøn
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 42,30 kr
    42,30 kr
    1 * 100 Items
       42,30 kr
  • BRAU4657624B
    Intravenous, intramuscular (size 12), made in china, with Max.shelf life 60 months
    Intravenous, intramuscular (size 12), made in china, with Max.shelf life 60 months
    22
    30 mm
    0,70 mm
    sort
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 57,10 kr
    57,10 kr
    1 * 100 Items
       57,10 kr