Orders LinkContactUsLinkComponent
Sampling swabs
Sampling swabs
Sampling swabs
Supplier:  DELTALAB
Certificates
Sampling swabs
4 Options Available Below
Product Details & Documents
For safe sampling. Item 115-8250 is longer than conventional swabs and was designed to reach hard-to-reach areas. For example, suitable for use in gynaecology when used with a speculum to remove endocervical cells. Also ideal for veterinary medicine.
  • All swabs have round, tight, tear-resistant heads to ensure perfect sampling without residues
Tip Ø: 5 mm
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Documents
Product Family Options
Product Information
  • Beskrivelse
    Skaft/hoved
    Ø
    Længde
    Steril
    Availability
    Price
  • Kun med træskaft
    Trae
    2,2 ±0,3 mm
    147 ±0,3 mm
    -
    Log in for availability
    Log in for availability
    1 * 1.000 Items
    1 * 1.000 Items - 214,00 kr
    214,00 kr
    1 * 1.000 Items
       214,00 kr
  • Sampling swabs
    Wood/cotton
    2,2 mm
    150 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 42,30 kr
    42,30 kr
    1 * 100 Items
       42,30 kr
  • Sampling swabs
    Wood/cotton
    2,5 mm
    200 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 136,00 kr
    136,00 kr
    1 * 100 Items
       136,00 kr
  • Sampling swabs
    PP/cotton
    2,5 mm
    200 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 46,40 kr
    46,40 kr
    1 * 100 Items
       46,40 kr