Orders LinkContactUsLinkComponent
Silikonepropper
Supplier:   HIRSCHMANN
Silikonepropper
Supplier:  HIRSCHMANN
Certificates
Silikonepropper
31 Options Available Below
Product Details & Documents
SILICOSEN® and BIO-SILICO® sterile stoppers are manufactured in a special process and have regular pores, thus making them ideal for the preparation and sterilisation of culture media. The uniform pores and good permeability to air make the stoppers ideally suitable for cultivation of aerobic microorganisms.
  • Chemical and temperature resistant
  • Liquid repellent
  • Reusable
SILICOSEN® T-type and L-type: Low humidity evaporation reduces dehydration of cultures.
SILICOSEN® C-type: Excellent permeability and secure fitting, thanks to seal lip. Ideally suitable for agitated cultures.
SILICOSEN® S-type: Excellent permeability and suitable for aerobic bacteria cultures and agitated cultures.
BIO-SILICO® N-type: High degree of air permeability, similar to that of cotton plugs.
Product Family Options
SILICOSEN® T-type
Product Information
  • Beskrivelse
    Farve
    Ant. huller
    Højde
    Ø bund
    Ø top
    Availability
    Price
  • SILICOSEN® T-type
    lyserød
    0
    30,0 mm
    6,0 mm
    10,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 277,00 kr
    277,00 kr
    1 * 10 Items
       277,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    30,0 mm
    9,0 mm
    12,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 326,00 kr
    326,00 kr
    1 * 10 Items
       326,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    35,0 mm
    11,0 mm
    15,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 385,00 kr
    385,00 kr
    1 * 10 Items
       385,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    40,0 mm
    13,0 mm
    17,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 423,00 kr
    423,00 kr
    1 * 10 Items
       423,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    42,0 mm
    15,0 mm
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 482,00 kr
    482,00 kr
    1 * 10 Items
       482,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    45,0 mm
    18,0 mm
    22,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 535,00 kr
    535,00 kr
    1 * 10 Items
       535,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    55,0 mm
    20,0 mm
    24,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 648,00 kr
    648,00 kr
    1 * 10 Items
       648,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    60,0 mm
    22,0 mm
    32,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 926,00 kr
    926,00 kr
    1 * 10 Items
       926,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    70,0 mm
    30,0 mm
    42,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 2.220,00 kr
    2.220,00 kr
    1 * 10 Items
       2.220,00 kr
  • SILICOSEN® T-type
    lyserød
    0
    100,0 mm
    40,0 mm
    52,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 3.530,00 kr
    3.530,00 kr
    1 * 10 Items
       3.530,00 kr
SILICOSEN® L-type
Product Information
  • Beskrivelse
    Farve
    Ant. huller
    Højde
    Ø bund
    Ø top
    Availability
    Price
  • SILICOSEN® L-type
    lyserød
    0
    50,0 mm
    9,0 mm
    12,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 482,00 kr
    482,00 kr
    1 * 10 Items
       482,00 kr
  • SILICOSEN® L-type
    lyserød
    0
    50,0 mm
    13,0 mm
    17,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 522,00 kr
    522,00 kr
    1 * 10 Items
       522,00 kr
  • SILICOSEN® L-type
    lyserød
    0
    60,0 mm
    18,0 mm
    22,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 847,00 kr
    847,00 kr
    1 * 10 Items
       847,00 kr
SILICOSEN® C-type
Product Information
  • Beskrivelse
    Farve
    Ant. huller
    Højde
    Ø bund
    Ø top
    Availability
    Price
  • SILICOSEN® C-type
    lyserød
    0
    28,0 mm
    12,0 mm
    16,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 792,00 kr
    792,00 kr
    1 * 10 Items
       792,00 kr
  • SILICOSEN® C-type
    lyserød
    0
    28,0 mm
    18,0 mm
    26,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 1.010,00 kr
    1.010,00 kr
    1 * 10 Items
       1.010,00 kr
  • SILICOSEN® C-type
    lyserød
    0
    28,0 mm
    27,0 mm
    36,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 1.380,00 kr
    1.380,00 kr
    1 * 10 Items
       1.380,00 kr
  • SILICOSEN® C-type
    lyserød
    0
    28,0 mm
    34,0 mm
    50,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 1.880,00 kr
    1.880,00 kr
    1 * 10 Items
       1.880,00 kr
  • SILICOSEN® C-type
    lyserød
    0
    29,0 mm
    42,0 mm
    57,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 2.330,00 kr
    2.330,00 kr
    1 * 10 Items
       2.330,00 kr
SILICOSEN® S-type
Product Information
  • Beskrivelse
    Farve
    Ant. huller
    Højde
    Ø bund
    Ø top
    Availability
    Price
  • SILICOSEN® S-type
    grøn
    0
    45,0 mm
    17,0 mm
    28,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 1.620,00 kr
    1.620,00 kr
    1 * 10 Items
       1.620,00 kr
  • SILICOSEN® S-type
    grøn
    0
    50,0 mm
    24,0 mm
    35,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 1.880,00 kr
    1.880,00 kr
    1 * 10 Items
       1.880,00 kr
  • SILICOSEN® S-type
    grøn
    0
    50,0 mm
    28,0 mm
    40,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 2.410,00 kr
    2.410,00 kr
    1 * 10 Items
       2.410,00 kr
BIO-SILICO® N-type
Product Information
  • Beskrivelse
    Farve
    Ant. huller
    Højde
    Ø bund
    Ø top
    Availability
    Price
  • BIO-SILICO® N-type
    orange
    0
    30,0 mm
    9,0 mm
    13,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 420,00 kr
    420,00 kr
    1 * 10 Items
       420,00 kr
  • BIO-SILICO® N-type
    orange
    0
    36,0 mm
    11,0 mm
    16,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 452,00 kr
    452,00 kr
    1 * 10 Items
       452,00 kr
  • BIO-SILICO® N-type
    orange
    0
    40,0 mm
    13,0 mm
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 487,00 kr
    487,00 kr
    1 * 10 Items
       487,00 kr
  • BIO-SILICO® N-type
    orange
    0
    44,0 mm
    15,0 mm
    21,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 529,00 kr
    529,00 kr
    1 * 10 Items
       529,00 kr
  • BIO-SILICO® N-type
    orange
    0
    48,0 mm
    18,0 mm
    24,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 634,00 kr
    634,00 kr
    1 * 10 Items
       634,00 kr
  • BIO-SILICO® N-type
    orange
    0
    52,0 mm
    20,0 mm
    25,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 776,00 kr
    776,00 kr
    1 * 10 Items
       776,00 kr
  • BIO-SILICO® N-type
    orange
    0
    54,0 mm
    21,0 mm
    30,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 989,00 kr
    989,00 kr
    1 * 10 Items
       989,00 kr
  • BIO-SILICO® N-type
    orange
    0
    60,0 mm
    22,0 mm
    34,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 1.220,00 kr
    1.220,00 kr
    1 * 10 Items
       1.220,00 kr
  • BIO-SILICO® N-type
    orange
    0
    78,0 mm
    30,0 mm
    44,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 2.870,00 kr
    2.870,00 kr
    1 * 10 Items
       2.870,00 kr
  • BIO-SILICO® N-type
    orange
    0
    86,0 mm
    40,0 mm
    54,0 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 3.330,00 kr
    3.330,00 kr
    1 * 10 Items
       3.330,00 kr