Orders LinkContactUsLinkComponent
Cotton wool
Cotton wool roll, not interleaved
  391-2264
Cotton wool
  391-2264
Cotton wool
Cotton wool
Cotton wool roll, not interleaved
405,00 kr
1 * 500 g
These non-absorbent bleached cotton wool rolls are made of 100% cotton.
  • Soft
  • Minimum linting
  • Minimum neps
  • Beskrivelse:
    Cotton wool roll, not interleaved