Orders LinkContactUsLinkComponent
Splash heads, sloping, Quickfit®
Splash heads, sloping, Quickfit®
24/29 NS, 24/29 NS
Katalog # BIBBSH7/43
Leverandør:   SciLabware
Splash heads, sloping, Quickfit®
Katalog # BIBBSH7/43
Leverandør:  SciLabware
Splash heads, sloping, Quickfit®
Splash heads, sloping, Quickfit®
24/29 NS, 24/29 NS
Price
4.680,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Used in distillation assemblies to prevent raw liquid 'carrying over' from flask to condenser.
Specifikationer
  • Konus til kondenser:
    24/29 NS
  • Konus til kolbe:
    24/29 NS
  • Seq:
    06