Laboratory marker
Fiberpen, sort
Katalog # 631-0014
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Permanent marker for indelibly labelling microscope slides and embedding cassettes with a high contrast black ink. Ultra-fine tip. These laboratory markers do not dry out and have a long lifetime.
- Water resistant, odour-free and non toxic
- Resistant against most laboratory chemicals
- No pretreatment of surfaces necessary
Documents
Specifikationer
- Farve:sort
- Beskrivelse:Fiberpen