Measuring spoons, set
Sæt med 4 måleskeer
231-1320
: BOCHEM
Stainless steel 18/10. Practical and convenient set of four measuring spoons.
- Heat and chemical resistant
- Ringed set keeps all sizes together
: Each set includes 1,25 ml, 2,5 ml, 5 ml and 15 ml spoons.
- Beskrivelse:Sæt med 4 måleskeer