Weighing scoop
170 mm, 10 mm
Catalog # 611-1251
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Stainless steel. Ideal for use as weighing scoops, weighing out etc.
- Corrosion resistant
- Robust and convenient to use, autoclavable
- Easy to clean
- For a variety of uses
Specifications
- Bredde:10 mm
- Længde:170 mm