Orders LinkContactUsLinkComponent
Spoon spatula, Nuffield type
Spoon spatula, Nuffield type
140 mm
Katalog # 231-0170
Leverandør:   NICKEL ELECTRO
Spoon spatula, Nuffield type
Katalog # 231-0170
Leverandør:  NICKEL ELECTRO
Spoon spatula, Nuffield type
Spoon spatula, Nuffield type
140 mm
Price
192,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Stainless steel. Straight with box joint.
  • Excellent thermal resistance, autoclavable
  • Excellent chemical resistance
  • Quick and easy to clean
Specifikationer
  • Længde:
    140 mm