Orders LinkContactUsLinkComponent
Adhesive seals
   Bæredygtige muligheder
Leverandør:   Greiner Bio-One
Adhesive seals
Leverandør:  Greiner Bio-One
   Bæredygtige muligheder
Certifikater
Adhesive seals
1 tilgængelige muligheder nedenfor
Product Details & Documents
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
  • Free from RNase, DNase and human DNA
  • Non pyrogenic
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 736-0127
  • GREI676001BE
  • GREI676050UK
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • ISO 14001 (241002); ISO 50001 (241002)
Specifikationer
Product Family Options
Produktinformation
  • Optisk egenskab
    Steril
    Tilgængelighed
    Price
  • 736-0127
    EASYseal-klæbefilm, usteril, 80×140 mm
    EASYseal-klæbefilm, usteril, 80×140 mm
    Transparent
    -
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 412,00 kr
    412,00 kr
    1 * 100 Items
       412,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    736-0127
    Environmentally Preferable
    Miljøcertificering
  • GREI676001BE
    EASYseal adhesive film, non sterile, 80×140 mm
    EASYseal adhesive film, non sterile, 80×140 mm
    Transparent
    -
    Discontinued
    Specifikationer:
    More
    Specifications
    Cat. No.
    GREI676001BE
    Environmentally Preferable
    Miljøcertificering
  • GREI676050UK
    BREATHseal adhesive film, non sterile
    BREATHseal adhesive film, non sterile
    Transparent
    Discontinued
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    GREI676050UK
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)