Orders LinkContactUsLinkComponent
Automation tips for Tecan®, Axygen®
Automation tips for Tecan®, Axygen®
   Sustainable Options Available
Supplier:   Corning
Automation tips for Tecan®, Axygen®
Supplier:  Corning
   Sustainable Options Available
Certificates
Automation tips for Tecan®, Axygen®
36 Options Available Below
Product Details & Documents
PP, med eller uden filter, sterile eller ikke sterile.
  • Black tips are conductive for liquid level-sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Packaging: Packaged in plastic blister packs.
Caution: * Economy packed in cardboard wrapper.
Specifications for Product Family
Specifications
  • Catalog No:
  • 732-0977
  • 732-0981
  • 732-0972
  • 732-0979
  • 732-0969
  • 732-0968
  • 732-0976
  • 732-0962
  • 732-0960
  • 732-0966
  • 732-0978
  • 732-0975
  • 732-0982
  • 732-0974
  • 736-0488
  • 732-0970
  • 732-0971
  • 732-0980
  • AXYGTTF900CBKHTRS
  • 736-0485
  • 732-0961
  • 732-0963
  • 732-0983
  • 736-0451
  • 736-0452
  • 736-0455
  • 736-0447
  • 736-0453
  • 736-0454
  • 736-0449
  • 736-0450
  • 736-0525
  • 736-0523
  • 613-3814
  • 613-3813
  • 613-3882
  • 613-3812
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Genesis Freedom®, Freedom Evo®, Miniprep med LiHa (klar)
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Spits farge
    Til brug med
    Pakket
    Availability
    Price
  • TTF-20-C-HTR-S
    20 µl
    +
    +
    Transparent
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.840,00 kr
    2.840,00 kr
    1 * 2.304 Items
       2.840,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0977
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-50-C-HTR-S
    50 µl
    +
    +
    Transparent
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.700,00 kr
    2.700,00 kr
    1 * 2.304 Items
       2.700,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0981
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-50-C-HTR
    50 µl
    -
    -
    Transparent
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.100,00 kr
    2.100,00 kr
    1 * 2.304 Items
       2.100,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0972
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-200-C-HTR-S
    200 µl
    +
    +
    Transparent
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.840,00 kr
    2.840,00 kr
    1 * 2.304 Items
       2.840,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0979
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-200-C-HTR-S
    200 µl
    +
    -
    Transparent
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.290,00 kr
    2.290,00 kr
    1 * 2.304 Items
       2.290,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0969
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-200-C-HTR
    200 µl
    -
    -
    Transparent
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.100,00 kr
    2.100,00 kr
    1 * 2.304 Items
       2.100,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0968
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-1000-C-HTR-S
    1000 µl
    +
    +
    Transparent
    LiHA-1, -4, -8
    16 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 1.536 Items
    1 * 1.536 Items - 1.890,00 kr
    1.890,00 kr
    1 * 1.536 Items
       1.890,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0976
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-1000-C-HTR-S
    1000 µl
    +
    -
    Transparent
    LiHA-1, -4, -8
    16 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 1.536 Items
    1 * 1.536 Items - 1.530,00 kr
    1.530,00 kr
    1 * 1.536 Items
       1.530,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0962
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-1000-C-HTR
    1000 µl
    -
    -
    Transparent
    LiHA-1, -4, -8
    16 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 1.536 Items
    1 * 1.536 Items - 1.400,00 kr
    1.400,00 kr
    1 * 1.536 Items
       1.400,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0960
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
Genesis Freedom®, Freedom Evo®, Miniprep med LiHa (sort)
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Spits farge
    Til brug med
    Pakket
    Availability
    Price
  • TT-20-CBK-HTR
    20 µl
    -
    -
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.640,00 kr
    2.640,00 kr
    1 * 2.304 Items
       2.640,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0966
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-20-CBK-HTR-S
    20 µl
    +
    +
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 3.560,00 kr
    3.560,00 kr
    1 * 2.304 Items
       3.560,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0978
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-50-CBK-HTR-S
    50 µl
    +
    -
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 3.010,00 kr
    3.010,00 kr
    1 * 2.304 Items
       3.010,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0975
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-50-CBK-HTR-S
    50 µl
    +
    +
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 3.560,00 kr
    3.560,00 kr
    1 * 2.304 Items
       3.560,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0982
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-50-CBK-HTR
    50 µl
    -
    -
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.640,00 kr
    2.640,00 kr
    1 * 2.304 Items
       2.640,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0974
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-50-CBK-HTR-36*
    50 µl
    -
    -
    Black
    LiHA-1, -4, -8
    72 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 6.912 Items
    1 * 6.912 Items - 6.480,00 kr
    6.480,00 kr
    1 * 6.912 Items
       6.480,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    736-0488
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-200-CBK-HTR
    200 µl
    -
    -
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.540,00 kr
    2.540,00 kr
    1 * 2.304 Items
       2.540,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0970
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-200-CBK-HTR-S
    200 µl
    +
    -
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 2.830,00 kr
    2.830,00 kr
    1 * 2.304 Items
       2.830,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0971
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-200-CBK-HTR-S
    200 µl
    +
    +
    Black
    LiHA-1, -4, -8
    24 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 2.304 Items
    1 * 2.304 Items - 3.460,00 kr
    3.460,00 kr
    1 * 2.304 Items
       3.460,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0980
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-900-CBK-HTR-S
    900 µl
    +
    +
    Black
    LiHA-1, -4, -8
    Log in for availability
    Log in for availability
    1 * 1.536 Items
    1 * 1.536 Items - 2.040,00 kr
    2.040,00 kr
    1 * 1.536 Items
       2.040,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AXYGTTF900CBKHTRS
    Environmentally Preferable
    Miljøcertificering
  • TT-1000-CBK-HTR-24*
    1000 µl
    -
    -
    Black
    LiHA-1, -4, -8
    48 rack, 96 each
    Log in for availability
    Log in for availability
    1 * 4.608 Items
    1 * 4.608 Items - 4.160,00 kr
    4.160,00 kr
    1 * 4.608 Items
       4.160,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    736-0485
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-1000-CBK-HTR
    1000 µl
    -
    -
    Black
    LiHA-1, -4, -8
    16 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 1.536 Items
    1 * 1.536 Items - 1.630,00 kr
    1.630,00 kr
    1 * 1.536 Items
       1.630,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0961
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TT-1000-CBK-HTR-S
    1000 µl
    +
    -
    Black
    LiHA-1, -4, -8
    16 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 1.536 Items
    1 * 1.536 Items - 1.830,00 kr
    1.830,00 kr
    1 * 1.536 Items
       1.830,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0963
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • TTF-1000-CBK-HTR-S
    1000 µl
    +
    +
    Black
    LiHA-1, -4, -8
    16 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 1.536 Items
    1 * 1.536 Items - 2.560,00 kr
    2.560,00 kr
    1 * 1.536 Items
       2.560,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    732-0983
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
Freedom Evo®, TeMo, Aquarius (klar)
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Spits farge
    Til brug med
    Pakket
    Availability
    Price
  • EV-50-R
    50 µl
    -
    -
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 4.980,00 kr
    4.980,00 kr
    1 * 4.800 Items
       4.980,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0451
    Environmentally Preferable
    Miljøcertificering
  • EV-50-R-S
    50 µl
    +
    -
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 5.130,00 kr
    5.130,00 kr
    1 * 4.800 Items
       5.130,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0452
    Environmentally Preferable
    Miljøcertificering
  • EVF-50-R-S
    50 µl
    +
    +
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 5.680,00 kr
    5.680,00 kr
    1 * 4.800 Items
       5.680,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0455
    Environmentally Preferable
    Miljøcertificering
  • EV-100-R
    100 µl
    -
    -
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 4.860,00 kr
    4.860,00 kr
    1 * 4.800 Items
       4.860,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0447
    Environmentally Preferable
    Miljøcertificering
  • EVF-100-R-S
    100 µl
    +
    +
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 5.680,00 kr
    5.680,00 kr
    1 * 4.800 Items
       5.680,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0453
    Environmentally Preferable
    Miljøcertificering
  • EVF-180-R-S
    180 µl
    +
    +
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 5.680,00 kr
    5.680,00 kr
    1 * 4.800 Items
       5.680,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0454
    Environmentally Preferable
    Miljøcertificering
  • EV-200-R
    200 µl
    -
    -
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 4.980,00 kr
    4.980,00 kr
    1 * 4.800 Items
       4.980,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0449
    Environmentally Preferable
    Miljøcertificering
  • EV-200-R-S
    200 µl
    +
    -
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 5.130,00 kr
    5.130,00 kr
    1 * 4.800 Items
       5.130,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    736-0450
    Environmentally Preferable
    Miljøcertificering
  • EVF-205-WB-R-S (bred åbning)
    200 µl
    +
    +
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Log in for availability
    Log in for availability
    1 * 4.800 Items
    1 * 4.800 Items - 7.110,00 kr
    7.110,00 kr
    1 * 4.800 Items
       7.110,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    736-0525
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • EV-205-WB-R (bred åbning)
    200 µl
    -
    -
    Transparent
    MCA96 / HV96
    50 racks, 96 i hvert
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    736-0523
    Environmentally Preferable
    Miljøcertificering
Tecan® Evo MCA96 (klar)
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Spits farge
    Til brug med
    Pakket
    Availability
    Price
  • EV-50-NTR-S
    50 µl
    +
    -
    Transparent
    MCA96
    96/Wafer, 8 Wafers/Stack, 5 Stacks/Case
    Log in for availability
    Log in for availability
    1 * 3.840 Items
    1 * 3.840 Items - 3.090,00 kr
    3.090,00 kr
    1 * 3.840 Items
       3.090,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3814
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • EV-50-NTR
    50 µl
    -
    -
    Transparent
    MCA96
    96/Wafer, 8 Wafers/Stack, 5 Stacks/Case
    Log in for availability
    Log in for availability
    1 * 3.840 Items
    1 * 3.840 Items - 2.980,00 kr
    2.980,00 kr
    1 * 3.840 Items
       2.980,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3813
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • EV-200-NTR
    200 µl
    -
    -
    Transparent
    MCA96
    96/Wafer, 8 Wafers/Stack, 5 Stacks/Case
    Log in for availability
    Log in for availability
    1 * 3.840 Items
    1 * 3.840 Items - 2.980,00 kr
    2.980,00 kr
    1 * 3.840 Items
       2.980,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3882
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • EV-200-NTR-S
    200 µl
    +
    -
    Transparent
    MCA96
    96/Wafer, 8 Wafers/Stack, 5 Stacks/Case
    Log in for availability
    Log in for availability
    1 * 3.840 Items
    1 * 3.840 Items - 3.090,00 kr
    3.090,00 kr
    1 * 3.840 Items
       3.090,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3812
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning