Pipettespidser, universal, Axygen®
Supplier: Corning
About this item
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
: Fås løse eller i racks, sterile eller usterile versioner.
59 Options Available Below
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
: Fås løse eller i racks, sterile eller usterile versioner.
Specifications for Product Family
Specifications
- Catalog No:
- 613-0640
- 613-0641
- 613-0628
- 613-0642
- 613-1309
- 613-0643
- 613-0645
- 613-0644
- 613-0646
- 613-0647
- 613-0648
- 613-1341
- 613-0649
- 613-0650
- 613-0629
- 613-0651
- 613-1288
- 613-0901
- 613-1289
- 613-1290
- 613-1291
- 613-1292
- 613-1293
- 613-1297
- 615-0029
- 613-1299
- 613-1338
- 613-1342
- 613-1343
- 613-1340
- 613-0656
- 613-0657
- 613-0658
- 613-0659
- 613-3827
- 613-3828
- 613-3829
- 613-3830
- 613-3826
- AXYGTR-222-C-STK-S
- 613-0661
- 613-0666
- 613-0667
- 613-0668
- 613-0669
- 613-0670
- 613-0630
- 613-0965
- 613-1270
- 613-1271
- 613-1272
- 613-1276
- 613-1277
- 613-1320
- 613-1334
- 613-1336
- 613-0673
- 613-1263
- 613-1267
- 613-1269
- 613-1265
- 613-0663
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Miljøcertificering:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
MicroVolume 10 µl
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-0640Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0641Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0628Environmentally PreferableMiljøcertificering - Standard0,5 - 10 µl-TransparentStack Rack inkl. 5 racks, 96 pr. stk.Log in for availability1 * 480 Items228,00 krSpecifications:
More
Specifications
Cat. No.613-0642Environmentally PreferableMiljøcertificering - Standard0,5 - 10 µl+TransparentStack rack incl. 5 racks, 96 eachLog in for availability1 * 480 Items258,00 krSpecifications:
More
Specifications
Cat. No.613-1309Environmentally PreferableMiljøcertificering
MicroVolume 10 µl,forlængede
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-0643Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0645Environmentally PreferableMiljøcertificering - Extended0,5 - 10 µl-TransparentStack racks, 96 pr. stk.Log in for availability1 * 960 Items453,00 krSpecifications:
More
Specifications
Cat. No.613-0644Environmentally PreferableMiljøcertificering
Ultra-Micro 20 µl
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-0646Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0647Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0648Environmentally PreferableMiljøcertificering
200 µl, Eppendorf compatible
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Eppendorf, Maxymum Recovery®1 - 200 µl+Yellow10 racks, 96 eachLog in for availability1 * 960 Items724,00 krSpecifications:
More
Specifications
Cat. No.613-1341Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-1338Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-1342Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-1343Environmentally PreferableMiljøcertificering - Eppendorf, Maxymum Recovery®1 - 200 µl-Yellow10 racks, 96 eachLog in for availability1 * 960 Items567,00 krSpecifications:
More
Specifications
Cat. No.613-1340Environmentally PreferableMiljøcertificering
200 µl, afrundet spids
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-0649Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringI-RECs, ISO 14001 - ReynosaLav fremstillingspåvirkning - Specifications:
More
Specifications
Cat. No.613-0650Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0629Environmentally PreferableMiljøcertificering - Bevelled1 - 200 µl-TransparentStack Rack inkl. 5 racks, 96 pr. stk.Log in for availability1 * 480 Items208,00 krSpecifications:
More
Specifications
Cat. No.613-0651Environmentally PreferableMiljøcertificering - Bevelled1 - 200 µl+TransparentStack rack incl. 5 racks, 96 eachLog in for availability1 * 480 Items266,00 krSpecifications:
More
Specifications
Cat. No.613-1288Environmentally PreferableMiljøcertificering - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-0901Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringI-RECs, ISO 14001 - ReynosaLav fremstillingspåvirkning - Specifications:
More
Specifications
Cat. No.613-1289Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-1290Environmentally PreferableMiljøcertificering - Bevelled1 - 200 µl-YellowStack Rack inkl. 5 racks, 96 pr. stk.Log in for availability1 * 480 Items210,00 krSpecifications:
More
Specifications
Cat. No.613-1291Environmentally PreferableMiljøcertificering - Bevelled1 - 200 µl+YellowStack rack incl. 5 racks, 96 eachLog in for availability1 * 480 Items238,00 krSpecifications:
More
Specifications
Cat. No.613-1292Environmentally PreferableMiljøcertificering - Bevelled, wide bore (1,5 mm)1 - 200 µl-TransparentBulkLog in for availability1 * 1.000 Items227,00 krSpecifications:
More
Specifications
Cat. No.613-1293Environmentally PreferableMiljøcertificering - Bevelled, wide bore (1,5 mm)1 - 200 µl-Transparent10 racks, 96 eachLog in for availability1 * 960 Items526,00 krSpecifications:
More
Specifications
Cat. No.613-1297Environmentally PreferableMiljøcertificering - Bevelled, wide bore (1,5 mm)1 - 200 µl+Transparent10 racks, 96 eachLog in for availability1 * 960 Items550,00 krSpecifications:
More
Specifications
Cat. No.615-0029Environmentally PreferableMiljøcertificering
200 µl
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-1299Environmentally PreferableMiljøcertificering
200 µl, afrundet spids, gradueret (10, 50, 100 µl)
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-0656Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringI-RECs, ISO 14001 - ReynosaLav fremstillingspåvirkning - Bevelled, graduated1 - 200 µl-Transparent10 racks, 96 pr. stk.Log in for availability1 * 960 Items425,00 krSpecifications:
More
Specifications
Cat. No.613-0657Environmentally PreferableMiljøcertificering - Bevelled, graduated1 - 200 µl+Transparent10 racks, 96 pr. stk.Log in for availability1 * 960 Items490,00 krSpecifications:
More
Specifications
Cat. No.613-0658Environmentally PreferableMiljøcertificering - Bevelled, graduated1 - 200 µl-TransparentStack Rack inkl. 5 racks, 96 pr. stk.Log in for availability1 * 480 Items213,00 krSpecifications:
More
Specifications
Cat. No.613-0659Environmentally PreferableMiljøcertificering - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-3827Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringI-RECs, ISO 14001 - ReynosaLav fremstillingspåvirkning - Bevelled, graduated1 - 200 µl-Yellow10 racks, 96 eachLog in for availability1 * 960 Items484,00 krSpecifications:
More
Specifications
Cat. No.613-3828Environmentally PreferableMiljøcertificering - Bevelled, graduated1 - 200 µl+Yellow10 racks, 96 eachLog in for availability1 * 960 Items512,00 krSpecifications:
More
Specifications
Cat. No.613-3829Environmentally PreferableMiljøcertificering - Bevelled, graduated1 - 200 µl-YellowStack rack incl. 5 racks, 96 eachLog in for availability1 * 480 Items213,00 krSpecifications:
More
Specifications
Cat. No.613-3830Environmentally PreferableMiljøcertificering - Bevelled, graduated, Maxymum Recovery®1 - 200 µl+TransparentStack rack incl. 5 racks, 96 eachLog in for availability1 * 480 Items266,00 krSpecifications:
More
Specifications
Cat. No.613-3826Environmentally PreferableMiljøcertificering - Bevelled, graduated1 - 200 µl+TransparentStack rack incl. 5 racks, 96 eachLog in for availability1 * 4.800 Items2.380,00 krSpecifications:
More
Specifications
Cat. No.AXYGTR-222-C-STK-SEnvironmentally PreferableMiljøcertificering
300 µl
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-0666Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0667Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-0668Environmentally PreferableMiljøcertificering
1000 µl, afrundet spids
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-0669Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringI-RECs, ISO 14001 - ReynosaLav fremstillingspåvirkning - Bevelled100 - 1000 µl-Transparent10 racks, 100 pr. stk.Log in for availability1 * 1.000 Items583,00 krSpecifications:
More
Specifications
Cat. No.613-0670Environmentally PreferableMiljøcertificering - Bevelled100 - 1000 µl+Transparent10 racks, 100 pr. stk.Log in for availability1 * 1.000 Items649,00 krSpecifications:
More
Specifications
Cat. No.613-0630Environmentally PreferableMiljøcertificering - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-0965Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringI-RECs, ISO 14001 - ReynosaLav fremstillingspåvirkning - Specifications:
More
Specifications
Cat. No.613-1270Environmentally PreferableMiljøcertificering - Specifications:
More
Specifications
Cat. No.613-1271Environmentally PreferableMiljøcertificering
1000 µl, wide bore
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-1272Environmentally PreferableMiljøcertificering - Wide bore (2 mm)100 - 1000 µl-Transparent10 racks, 100 eachLog in for availability1 * 1.000 Items785,00 krSpecifications:
More
Specifications
Cat. No.613-1276Environmentally PreferableMiljøcertificering - Wide bore (2 mm)100 - 1000 µl+Transparent10 racks, 100 eachLog in for availability1 * 1.000 Items1.110,00 krSpecifications:
More
Specifications
Cat. No.613-1277Environmentally PreferableMiljøcertificering
1000 µl, Eppendorf
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-1320Environmentally PreferableMiljøcertificering - Eppendorf, graduated, Maxymum Recovery®100 - 1000 µl-Blue12 racks, 96 eachLog in for availability1 * 1.152 Items869,00 krSpecifications:
More
Specifications
Cat. No.613-1334Environmentally PreferableMiljøcertificering - Eppendorf, graduated100 - 1000 µl-Blue12 racks, 96 eachLog in for availability1 * 1.152 Items903,00 krSpecifications:
More
Specifications
Cat. No.613-1336Environmentally PreferableMiljøcertificering
1250 µl, afrundet spids
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.613-1263Environmentally PreferableMiljøcertificering - Bevelled, Matrix-style15 - 1250 µl-Transparent12 racks, 96 eachLog in for availability1 * 1.152 Items1.470,00 krSpecifications:
More
Specifications
Cat. No.613-1267Environmentally PreferableMiljøcertificering - Bevelled, Matrix-style15 - 1250 µl+Transparent12 racks, 96 eachLog in for availability1 * 1.152 Items1.790,00 krSpecifications:
More
Specifications
Cat. No.613-1269Environmentally PreferableMiljøcertificering - Bevelled, Maxymum Recovery®15 - 1250 µl-Transparent12 racks, 96 eachDiscontinued-Specifications:
More
Specifications
Cat. No.613-1265Environmentally PreferableMiljøcertificering
Makrovolumen 5000 µl
Product Information
- SpidstypeVolumenSterilSpits fargePakketAvailabilityPrice
- Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-0663Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringI-RECs, ISO 14001 - ReynosaLav fremstillingspåvirkning