Orders LinkContactUsLinkComponent
Transfer pipettes, paddle
Transfer pipettes, paddle
0,2 ml, 50 µl, 124, -, Pose
Katalog # 612-1731
Leverandør:   Alpha Laboratories
Transfer pipettes, paddle
Katalog # 612-1731
Leverandør:  Alpha Laboratories
Transfer pipettes, paddle
Transfer pipettes, paddle
0,2 ml, 50 µl, 124, -, Pose
Price
624,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Non toxic, food grade LDPE.
  • For dispensing and mixing applications and popular with diagnostic kit manufacturers
  • Ideal for latex agglutination, pregnancy tests and blood grouping cards
Specifikationer
  • Pakket:
    Pose
  • Steril:
    -
  • Kapacitet:
    0,2 ml
  • Længde:
    124
  • Dråbevol.:
    50 µl