Pipettehjælper, pipetus®-junior
Catalog # 612-2604
Supplier: HIRSCHMANN
Specifications
- Beskrivelse:pipetus®-junior, komplet
- Pakn:1
Specifications
About this item
Pipette controller for glass and plastic pipettes from 0,5 to 25 ml.
- Easy, ergonomic, safe and accurate
- Total expulsion of the liquid simply by pressing the button on the back of the instrument
- The integrated, hydrophobic PTFE membrane filter ensures the pipette is protected against accidental overfilling
Full pressure on the fill button allows suction of around 2 ml. By pressing on the empty button, the pipette is emptied gravimetrically and the degree of pressure on the button determines the rate of flow.