Orders LinkContactUsLinkComponent
Mop systems, Edgeless®
Mop systems, Edgeless®
Leverandør:   CONTEC
Mop systems, Edgeless®
Leverandør:  CONTEC
Certifikater
Mop systems, Edgeless®
Mop systems, Edgeless®
3 tilgængelige muligheder nedenfor
Product Details & Documents
Designed for wet mopping in controlled environments and cleanrooms for effective cleaning and retrieval of excess fluid. Ideal for use in sterile environments. Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption.
  • Mop heads are free from any cellulose contaminants
  • Stainless bucket, downpress wringer and insert maximise wringing pressure and effectiveness
  • All system components and buckets are compatible with gamma, ETO, and autoclave sterilisation
Moppehovederne vaskes og pakkes i et ISO klasse 4 (FED-STD-209E klasse 10/M2,5) rentrum.
Bestillingsoplysninger: Alle moppesystemkomponenter skal bestilles separat. Plastspandssystemer inkluderer vogn, spande, vrider og styrehjul. Spandsystem af rustfrit stål inkluderer vogn spande, vrider og styrehjul.
Product Family Options
Produktinformation
  • Dimensioner
    Længde
    Kapacitet
    Tilgængelighed
    Price
  • CONTEL-MOPIR
    Standard edgeless mop head, sterile
    Standard edgeless mop head, sterile
    -
    -
    -
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 20 Items
    1 * 20 Items - kr10,560.00
    kr10,560.00
    1 * 20 Items
       kr10,560.00
  • 115-0174
    Standard moppehoved u. kanter, usterilt
    Standard moppehoved u. kanter, usterilt
    -
    -
    -
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr465.00
    kr465.00
    1 * 1 Items
       kr465.00
  • CONT2646F
    Edgeless floor mop frame QuickConnect™, polypropylene copolymer frame with stainless steel connector ; compatible with all styles of Edgeless mops and easily attaches to QuickConnect handles. Autoclavable at 121°C for 30 min.
    Edgeless floor mop frame QuickConnect™, polypropylene copolymer frame with stainless steel connector ; compatible with all styles of Edgeless mops and easily attaches to QuickConnect handles. Autoclavable at 121°C for 30 min.
    150×180 mm
    -
    -
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr1,450.00
    kr1,450.00
    1 * 1 Items
       kr1,450.00